Skip to Content
Merck
All Photos(1)

Key Documents

Y0000326

Azelastine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)phthalazin-1-one hydrochloride, Astelin, Optivar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24ClN3O · HCl
CAS Number:
Molecular Weight:
418.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

azelastine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN1CCCC(CC1)N2N=C(Cc3ccc(Cl)cc3)c4ccccc4C2=O

InChI

1S/C22H24ClN3O.ClH/c1-25-13-4-5-18(12-14-25)26-22(27)20-7-3-2-6-19(20)21(24-26)15-16-8-10-17(23)11-9-16;/h2-3,6-11,18H,4-5,12-15H2,1H3;1H

InChI key

YEJAJYAHJQIWNU-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azelastine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

H1 histamine receptor antagonist; NF-kB activator.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eli O Meltzer et al.
Allergy and asthma proceedings, 33(4), 324-332 (2012-08-04)
Many patients with allergic rhinitis (AR) have uncontrolled symptoms despite available treatment options. This study was designed to evaluate the efficacy and safety of MP29-02 (a novel intranasal formulation of fluticasone propionate [FP] and azelastine [AZ] hydrochloride), compared with monotherapy
Tsugunobu Andoh et al.
Journal of pharmacological sciences, 114(3), 292-297 (2010-10-16)
Recently, we showed that a methanol extract of Ganoderma lucidum inhibits scratching, an itch-related response, induced by intradermal injections of some pruritogens in mice. The present study investigated whether G. lucidum extract would inhibit allergic itch. In mice sensitized with
Carsten Gründemann et al.
Journal of ethnopharmacology, 145(1), 118-126 (2012-11-13)
Extracts from Veronica officinalis L. are traditionally used for the treatment of lung diseases; however, the effective compounds and the mode of action are still unknown. Here we analyzed the effects of a standardized Veronica extract on genes expression and
Jonathan A Bernstein
Current medical research and opinion, 23(10), 2441-2452 (2007-08-29)
Azelastine hydrochloride (Astelin) nasal spray 0.1% solution is a second-generation intranasal antihistamine available in the US for treatment of both seasonal allergic rhinitis (SAR) and nonallergic vasomotor rhinitis (VMR). Searches of journal articles including the title word 'azelastine' from 1979
Hartmut Derendorf et al.
British journal of clinical pharmacology, 74(1), 125-133 (2012-02-24)
• The topical second generation anti-histamine azelastine hydrochloride (AZE) and the potent corticosteroid fluticasone propionate (FP) are well established first-line treatments in allergic rhinitis (AR). • MP29-02, a novel intranasal AZE and FP formulation, has been shown to control AR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service