Skip to Content
Merck
All Photos(1)

Key Documents

Y0000109

Thimerosal

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thimerosal

manufacturer/tradename

EDQM

mp

234-237 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thimerosal EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Disclaimer

As part of the corporation global plan to support the Minamata Convention in eliminating mercury-containing compounds, as of January 2021 customers will be required to provide an End-Use Declaration. The sales of thimerosal will be restricted for approved applications only, such as preservative in vaccine manufacturing, preservative in eye cosmetics, research use within academia (excluding preservation intent) and as reference material only.

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act , European Biocidal Products Regulation, Canada′s Pest Management Regulatory Agency, Turkey′s Biocidal Products Regulation, Korea′s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Magos
Journal of applied toxicology : JAT, 23(4), 263-269 (2003-07-29)
The decomposition rate of organomercurials and the potency of the blood-brain barrier increase with the size of the organic radical. Thus methylmercury damages the brain more than thimerosal does, and when intake limits set for methylmercury are applied to thimerosal
Sarah K Parker et al.
Pediatrics, 114(3), 793-804 (2004-09-03)
The issue of thimerosal-containing vaccines as a possible cause of autistic spectrum disorders (ASD) and neurodevelopmental disorders (NDDs) has been a controversial topic since 1999. Although most practitioners are familiar with the controversy, many are not familiar with the type
H Möller
Contact dermatitis, 31(4), 209-213 (1994-10-01)
By and large, positive patch tests with thimerosal lack clinical relevance and should cause concern neither to the dermatologist nor to the patient. The substance should be retained in standard test series for scientific purposes, but on clinical grounds its
J G Elferink
General pharmacology, 33(1), 1-6 (1999-07-31)
An overview of the literature concerning the effects of thimerosal is presented. Because of its antibacterial effect, thimerosal is used for a variety of practical purposes such as antiseptic and preservative. In biomedical studies, thimerosal is used as a sulfhydryl
Vaccines, thimerosal, and neurodevelopmental outcomes.
Lawrence M Herman et al.
JAAPA : official journal of the American Academy of Physician Assistants, 19(1), 16-16 (2006-01-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service