Skip to Content
Merck
All Photos(1)

Key Documents

X405

Sigma-Aldrich

Triton X-405 solution

70% in H2O

Synonym(s):

4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol solution, Polyethylene glycol tert-octylphenyl ether, Polyoxyethylene (40) isooctylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

Quality Level

mol wt

~1967.0 g/mol

concentration

70% in H2O

technique(s)

ELISA: suitable

CMC

0.81 mM

transition temp

cloud point >100 °C

density

1.096 g/mL at 25 °C

HLB

17.6

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-405 comprises a longer chain of -CH2-CH2-O- monomers.

Application

Triton X-405 solution has been used:
  • to re-disperse semiconducting single-walled carbon nanotubes (S-SWCNTs/SWCNTs) for absorption spectroscopic characterization
  • as a component of sodium hydroxide solution to solubilize animal carcasses for liquid scintillation counting
  • as a surfactant in direct foaming to study its effects on the viscosity of geopolymer pastes

Caution

May precipitate on storage; redissolve at 50 °C

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Athersuch et al.
Xenobiotica; the fate of foreign compounds in biological systems, 40(7), 510-523 (2010-05-07)
1. A study of the rates and routes of excretion of 3-fluoro-[U-(14)C]aniline following intraperitoneal administration to male bile-cannulated rats by liquid scintillation counting (LSC) gave a total recovery of approximately 90% in the 48 h following dosing, with the majority
Li Wei et al.
Nanoscale, 9(32), 11640-11646 (2017-08-05)
Ultrahigh purity semiconducting single-walled carbon nanotubes (S-SWCNTs) are required for high-performance transistors. Aqueous two-phase (ATP) separation is an attractive method to obtain such SWCNTs due to its simplicity and scalability. This work targeted two questions; namely what is the upper
Silke Leutner et al.
PLoS neglected tropical diseases, 7(11), e2532-e2532 (2013-11-19)
Schistosomiasis is a disease of world-wide importance and is caused by parasitic flatworms of the genus Schistosoma. These parasites exhibit a unique reproduction biology as the female's sexual maturation depends on a constant pairing-contact to the male. Pairing leads to
Steffen Hahnel et al.
PLoS neglected tropical diseases, 7(7), e2336-e2336 (2013-08-13)
Schistosomiasis is one of the most important parasitic diseases worldwide, second only to malaria. Schistosomes exhibit an exceptional reproductive biology since the sexual maturation of the female, which includes the differentiation of the reproductive organs, is controlled by pairing. Pathogenicity
Stephanie van de Wall et al.
Vaccines, 3(2), 221-238 (2015-09-08)
The skin is an attractive organ for immunization because of the presence of antigen-presenting cells. Intradermal delivery via tattooing has demonstrated superior vaccine immunogenicity of DNA vaccines in comparison to conventional delivery methods. In this study, we explored the efficacy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service