Skip to Content
Merck
All Photos(1)

Key Documents

VP001

SAFC

VirusExpress® 293T Lentiviral Production Cells

Pharma Manufacturing

Synonym(s):

HEK293T cells for lentivirus production

Sign Into View Organizational & Contract Pricing


About This Item

shipped in

liquid nitrogen

Quality Level

storage temp.

−196°C

General description

VirusExpress 293T Lentiviral Production Cells are a clonal, suspension-adapted HEK293T derivative selected for fast growth, high transfection efficiency, and high-titer lentivirus production. VirusExpress 293T Lentiviral Production Cells grow to high cell densities, exhibit minimal cell clumping, and are amenable to PEI-based transfection. A medium exchange step is not needed prior to transfection with a PEI transfection reagent, and the cells can be transfected in larger volumes, enabling scale-up of lentivirus production. The cells are intended to be cultured in, and can be directly thawed into EX-CELL® CD HEK293 Viral Vector Medium.

VirusExpress 293T Lentiviral Production Cells are derived from the HEK293T cell line (ATCC CRL-3216) through single cell cloning. The parental HEK293 cell line was established by transformation of human embryonic kidney cells with sheared human adenovirus 5 DNA (Graham FL et al. 1977). A 4-kb adenoviral genome fragment is known to have integrated into chromosome 19 and encodes for E1A/E1B proteins. HEK293T cells were created by stable transfection of the HEK293 cell line with a temperature-sensitive mutant of SV40 large T antigen. Due to the expression of the large T antigen, transfected plasmids that carry the SV40 origin of replication can replicate in 293T cells and will transiently maintain a high copy number. Our clonal derivative eliminates variability from the population while also being adapted to suspension, serum-free conditions. Suspension culture and chemically defined medium allow scalability into stirred tank bioreactors for yields that can meet commercial needs.

Packaging

This product is shipped in a liquid nitrogen dry vapor shipper. The liquid nitrogen dry vapor shipper is not disposable and should be returned after receipt.

Other Notes

The cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL. The cells were banked using under cGMP conditions in chemically defined medium supplement with 6 mM glutamine and 8% DMSO.

Legal Information

A commercial license must be taken prior to use of the cells, or any products or materials derived from the cells, in a manufacturing process. Please contact your local sales representative for more details.
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
VirusExpress is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn how development of a suspension adapted HEK293T cell line with accompanying cell culture media can maximize lentivirus production for cell and gene therapies.

Learn how development of a suspension adapted HEK293T cell line with accompanying cell culture media can maximize lentivirus production for cell and gene therapies.

Learn how development of a suspension adapted HEK293T cell line with accompanying cell culture media can maximize lentivirus production for cell and gene therapies.

Learn how development of a suspension adapted HEK293T cell line with accompanying cell culture media can maximize lentivirus production for cell and gene therapies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service