Skip to Content
Merck
All Photos(5)

Key Documents

T7875

Sigma-Aldrich

Thiourea

ReagentPlus®, ≥99.0%

Synonym(s):

Sulfourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Beilstein:
605327
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

mp

170-176 °C (lit.)

functional group

amine
thiourea

SMILES string

NC(N)=S

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Gene Information

mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

General description

Thiourea is an analogue of urea, containing sulphur.[1]

Application

Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins
Thiourea has been used:
  • as a component in ice cold sample buffer for protein analysis [2]
  • in intracellular extraction buffer for the extraction of soluble intracellular proteins[3]
  • as a component of electrophoresis buffer[4]

Biochem/physiol Actions

Thiourea is a free radical scavenger of the peroxide radical.[5] It was shown to inhibit lipid peroxidation and ultraviolet (UV)-induced crosslinking of collagen. Bud dormancy in plants can be inhibited by thiourea, which is used as a growth stimulator. It is also known to be used in the treatment of hyperthyroidism.[1]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synergistic effects of glucose and ultraviolet irradiation on the physical properties of collagen
Ohan MP, et al.
Journal of Biomedical Materials Research, 60(3), 384-391 (2002)
Feline hypertrophic cardiomyopathy associated with the p. A31P Mutation in cMyBP-C is caused by production of mutated cMyBP-C with reduced binding to actin
Godiksen MTN, et al.
Open Journal of Veterinary Medicine, 3(02), 95-95 (2013)
Extraction of Intracellular and Cell Wall Proteins from Leaves and Roots of Harsh Hakea
Shane MW and Plaxton WC
Journal of Experimental Botany (2014)
Protocol: Optimised methodology for isolation of nuclei from leaves of species in the Solanaceae and Rosaceae families
Sikorskaite S, et al.
Plant methods, 9(1), 31-31 (2013)
Mechanism of the reduction of methylene blue by thiourea in aqueous acidic medium
Osunlaja AA
International Journal of ChemTech Research, 4(2), 609-617 (2012)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service