T59455
Triethylene glycol
ReagentPlus®, 99%
Synonym(s):
Triglycol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
150.17
Beilstein:
969357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
bp:
125-127 °C/0.1 mmHg (lit.)
vapor pressure:
<0.01 mmHg ( 20 °C)
Recommended Products
vapor density
5.2 (vs air)
Quality Level
vapor pressure
<0.01 mmHg ( 20 °C)
product line
ReagentPlus®
Assay
99%
form
viscous liquid
autoignition temp.
699 °F
expl. lim.
9.2 %
refractive index
n20/D 1.455 (lit.)
pH
6.5-7.5 (20 °C, 100 g/L)
bp
125-127 °C/0.1 mmHg (lit.)
mp
−7 °C (lit.)
density
1.124 g/mL at 20 °C (lit.)
functional group
ether
hydroxyl
SMILES string
OCCOCCOCCO
InChI
1S/C6H14O4/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2
InChI key
ZIBGPFATKBEMQZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Triethylene glycol can be used:
- To prepare fatty acid gelators, which are used to gelate various edible and vegetable oils.
- As a solvent to prepare superparamagnetic iron oxide nanoparticles for in situ protein purification.
- As an absorbent agent in the subsea natural gas dehydration process.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
330.8 °F - closed cup
Flash Point(C)
166 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Design and physicochemical properties of long and stiff fatty low molecular weight oleogelators
Delbecq F, et al.
Journal of Molecular Liquids, 295, 111708-111708 (2019)
Yeong-Ji Jang et al.
Diagnostics (Basel, Switzerland), 11(1) (2020-12-31)
The water proton spin relaxivity, colloidal stability, and biocompatibility of nanoparticle-based magnetic resonance imaging (MRI) contrast agents depend on the surface-coating ligands. Here, poly(acrylic acid-co-maleic acid) (PAAMA) (Mw = ~3000 amu) is explored as a surface-coating ligand of ultrasmall gadolinium
Mohammad Yaseen Ahmad et al.
Molecules (Basel, Switzerland), 25(5) (2020-03-11)
The study of ultra-small paramagnetic gadolinium oxide (Gd2O3) nanoparticles (NPs) as in vivo positive (T1) magnetic resonance imaging (MRI) contrast agents is one of the most attractive fields in nanomedicine. The performance of the Gd2O3 NP imaging agents depends on
Subsea natural gas dehydration in a membrane contactor with turbulence promoter: An experimental and modeling study
Ahmadi M, et al.
Chemical Engineering Journal, 404(15), 126535-126535 (2021)
Spray-Dried Hierarchical Aggregates of Iron Oxide Nanoparticles and Their Functionalization for Downstream Processing in Biotechnology
Kleinfeldt L, et al.
ACS Omega, 4(15), 16300-16308 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service