Skip to Content
Merck
All Photos(1)

Key Documents

T2100000

Trimethadione

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3,5,5,-Trimethyloxazolidine-2,4-dione, Trimethadione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO3
CAS Number:
Molecular Weight:
143.14
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trimethadione

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1C(=O)OC(C)(C)C1=O

InChI

1S/C6H9NO3/c1-6(2)4(8)7(3)5(9)10-6/h1-3H3

InChI key

IRYJRGCIQBGHIV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trimethadione EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Tanaka et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 104(2), 205-207 (1993-02-01)
1. Trimethadione (TMO) has the properties required of probe drugs for the evaluation of hepatic oxidizing capacity in vivo. 2. TMO is demethylated to dimethadione (DMO), its only metabolite, in the liver after oral administration. 3. In rats with various
[Fetal trimethadione effects].
Y Nakane
Ryoikibetsu shokogun shirizu, (30 Pt 5)(30 Pt 5), 94-95 (2000-11-01)
[Fetal trimethadione syndrome].
T Nagai
Ryoikibetsu shokogun shirizu, (33)(33), 704-704 (2001-07-21)
E Tanaka et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 115(3), 211-216 (1996-11-01)
Trimethadione (TMO) has the properties required of probe drugs for the evaluation of hepatic drug-oxidizing capacity in humans in vivo. TMO is demethylated to dimethadione (DMO), its only metabolite, in the liver after oral administration. Involvement of two cytochrome P450's--CYP2C9

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service