Skip to Content
Merck
All Photos(1)

Documents

T1950000

Triamcinolone hexacetonide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(11β,16α)-21-(3,3-Dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H41FO7
CAS Number:
Molecular Weight:
532.64
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triamcinolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C30H41FO7/c1-25(2,3)15-24(35)36-16-22(34)30-23(37-26(4,5)38-30)13-20-19-9-8-17-12-18(32)10-11-27(17,6)29(19,31)21(33)14-28(20,30)7/h10-12,19-21,23,33H,8-9,13-16H2,1-7H3/t19-,20-,21-,23+,27-,28-,29-,30+/m0/s1

InChI key

TZIZWYVVGLXXFV-FLRHRWPCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triamcinolone hexacetonide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leandro Parmigiani et al.
Clinical rheumatology, 29(11), 1311-1315 (2010-07-14)
Compare the medium-term effectiveness and tolerance between joint lavage (JL) in combination with triamcinolone hexacetonide (TH) intra-articular injection (IAI) and IAI with TH alone for treatment of primary osteoarthritis (OA) of the knee. A randomized, double-blind, controlled study was carried
B Anne Eberhard et al.
The Journal of rheumatology, 39(2), 374-376 (2011-12-03)
To determine whether the intraarticular (IA) dose of triamcinolone hexacetonide (TH) or triamcinolone acetonide (TA) influences time to relapse among patients with juvenile idiopathic arthritis (JIA). The primary endpoint variable was the time to relapse of arthritis in the affected
R V Lopes et al.
Rheumatology (Oxford, England), 47(12), 1792-1794 (2008-09-30)
To assess the accuracy of blind IA injections (IAIs) in patients with RA and determine their effectiveness. A controlled prospective study including 96 RA patients undergoing blind IAIs in peripheral joints was carried out. Clinical evaluation was performed at baseline
Marla Francisca dos Santos et al.
Clinics (Sao Paulo, Brazil), 64(12), 1187-1193 (2009-12-29)
The aim of the present study was to investigate the effectiveness of Samarium(153)-particulate hydroxyapatite radiation synovectomy in rheumatoid arthritis patients with chronic knee synovitis. Fifty-eight rheumatoid arthritis patients (60 knees) with chronic knee synovitis participated in a controlled double-blinded trial.
Matthew L Stoll et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 70(8), 1802-1807 (2012-01-24)
The purpose of this study was to evaluate the safety and efficacy of intra-articular corticosteroid injections (IACIs) of the temporomandibular joint (TMJ) in children with juvenile idiopathic arthritis (JIA) when administered by an oral and maxillofacial surgeon without imaging guidance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service