Skip to Content
Merck
All Photos(1)

Key Documents

T1410900

Tiaprofenic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Tiaprofenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3S
CAS Number:
Molecular Weight:
260.31
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tiaprofenic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[s]1c(ccc1C(=O)c2ccccc2)C(C)C(=O)O

InChI

1S/C14H12O3S/c1-9(14(16)17)11-7-8-12(18-11)13(15)10-5-3-2-4-6-10/h2-9H,1H3,(H,16,17)

InChI key

GUHPRPJDBZHYCJ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tiaprofenic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariko Sugiura et al.
Photodermatology, photoimmunology & photomedicine, 18(2), 82-89 (2002-07-31)
Ketoprofen, suprofen and tiaprofenic acid are arylpropionic anti-inflammatories. Their chemical structures share the same elements as the benzoyl radical and the tiophene ring. We experienced nine cases of ketoprofen photoallergy, seven cases of suprofen photoallergy and three cases of tiaprofenic
[Tuberculous osteitis of the malar bone].
G Ondzotto
Medecine tropicale : revue du Corps de sante colonial, 62(5), 564-565 (2003-03-06)
U Tursen et al.
The Journal of dermatological treatment, 13(4), 205-206 (2002-12-01)
Tiaprofenic acid is a propionic acid derivative with analgesic, anti-inflammatory and antipyretic properties. Fixed drug eruption is a common cutaneous reaction by various drugs. Fixed drug eruption induced by tiaprofenic acid has not been reported previously. Described in this paper
A Samadi et al.
Photochemistry and photobiology, 73(4), 359-365 (2001-05-03)
A careful study of the linoleic acid hydroperoxide (LOOH) profile obtained upon peroxidation of linoleic acid (LA) photosensitized by tiaprofenic acid (TPA) and analogous ketones has been undertaken to distinguish between type-I and type-II photoperoxidation mechanisms. 1,4-Cyclohexadiene and 1,2-dimethylcyclohexa-2,5-dienecarboxylic acid
F Boscá et al.
Journal of photochemistry and photobiology. B, Biology, 58(1), 1-5 (2001-02-24)
Analysis of the photomixtures resulting from irradiation of aqueous solutions of linoleic acid sensitized by tiaprofenic acid (TPA) or its major photoproduct (DTPA) by HPLC has shown the formation of all the four possible conjugated dienic hydroperoxides. According to laser

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service