Skip to Content
Merck
All Photos(4)

Documents

S7907

Sigma-Aldrich

Sodium phosphate dibasic

BioXtra, ≥99.0%

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.0%

form

powder or crystals

impurities

≤0.1% Insoluble matter

pH

8.0-11.0 (20 °C, 1 M in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.002%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

absorption

<0.05 at 260 in H2O at 1 M
<0.05 at 280 in H2O at 1 M

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate dibasic (NaH2PO4), also known as disodium phosphate, is a commonly used phosphate salt in biochemistry research due to its versatile properties. It is a key component in the preparation of phosphate buffer saline (PBS) and phosphate buffer, a commonly used buffer solution in many biological applications. In addition to its buffer properties, NaH2PO4 is also used as a precipitating agent to form silver orthophosphate (Ag3PO4) powders, making it a valuable tool in many laboratory settings. Disodium phosphate is also a widely used emulsifier in dairy products such as cheese and other foods.

Application

Sodium phosphate dibasic has been used:
  • as a component of phosphate buffered saline (PBS)
  • as a component of buffer for the preparation of cryopreservation diluent
  • in the preparation of McIlvaine buffer (citrate-phosphate buffer) for toluidine blue staining
  • in the preparation of the S9 mix for the Ames test, and SOS Chromotest
  • in the preparation of calcium deficient hydroxyapatite (CDHA) paste during the fabrication process of the CDHA scaffold

Features and Benefits

  • The BioXtra product line ensures high-quality products for consistent results.
  • High level of assay ( ≥99.0%), indicating its purity.
  • Powder form for easy handling and use.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sperm protamine-status correlates to the fertility of breeding bulls
Dogan S, et al.
Biology of Reproduction, 92(4) (2015)
Effect of Emulsifier Salts on Textural and Flavor Properties of Processed Cheeses
Gupta SK, et al.
Journal of Dairy Science, 67(4), 764-778 (1984)
Sequential generations of honey bee (Apis mellifera) queens produced using cryopreserved semen
Hopkins BK, et al.
Reproduction, Fertility, and Development, 24(8), 1079-1083 (2012)
Injectable magnesium oxychloride cement foam-derived scaffold for augmenting osteoporotic defect repair
Zhu Y, et al.
Journal of Colloid and Interface Science (2023)
Whole-mount Immunohistochemistry of Adult Zebrafish Retina for Advanced Imaging
Nagashima M and Hitchcock PF
Bio-protocol, 10(24) (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service