Skip to Content
Merck
All Photos(1)

Key Documents

S2190000

Sulpiride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Sulpiride, (±)-5-(Aminosulfonyl)-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide, (±)-N-1-(Ethylpyrrolidin-2-ylmethyl)-2-methoxy-5-sulfamoylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N3O4S
CAS Number:
Molecular Weight:
341.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulpiride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN1CCCC1CNC(=O)c2cc(ccc2OC)S(N)(=O)=O

InChI

1S/C15H23N3O4S/c1-3-18-8-4-5-11(18)10-17-15(19)13-9-12(23(16,20)21)6-7-14(13)22-2/h6-7,9,11H,3-5,8,10H2,1-2H3,(H,17,19)(H2,16,20,21)

InChI key

BGRJTUBHPOOWDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulpiride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J H Stubbs et al.
Acta psychiatrica Scandinavica, 102(5), 390-393 (2000-12-01)
Schizophrenic patients who are only partially responsive to clozapine pose a therapeutic challenge. In these circumstances some clinicians would consider adding in a second antipsychotic. We present a case report and review evidence for the efficacy of such augmentation strategies.
G Serra et al.
Clinical neuropharmacology, 13 Suppl 1, S76-S83 (1990-01-01)
L-Sulpiride is a dopamine (DA)-receptor blocker that acts as an antidepressant at low doses. We have reviewed evidence suggesting that the antidepressant effect of L-sulpiride may depend on the activation of DA transmission secondary to the blockade of DA autoreceptors.
Sulpiride augmentation for schizophrenia.
Jijun Wang et al.
Schizophrenia bulletin, 36(2), 229-230 (2010-01-12)
P Jenner et al.
Advances in biochemical psychopharmacology, 32, 85-103 (1982-01-01)
It can be said that sulpiride exerts a disinhibitory effect in both depression and schizophrenia but this is not associated with mechanisms through which typical antidepressant or anxiolytic agents act. Sulpiride acts selectively as a dopamine receptor antagonist in the
K R Müller-Vahl
Der Nervenarzt, 78(3), 264-264 (2006-08-23)
The treatment of Tourette's syndrome is a challenge. Dopamine receptor antagonists are the drugs of first choice for the treatment of tics. Because large controlled trials are lacking, there is no consensus about which of the different neuroleptic drugs should

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service