Skip to Content
Merck
All Photos(1)

Key Documents

S2159000

Sulfinpyrazone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Sulfinpyrazone, 1,2-Diphenyl-4-(phenylsulfinylethyl)-3,5-pyrazolidinedione, Diphenylpyrazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N2O3S
CAS Number:
Molecular Weight:
404.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfinpyrazone

manufacturer/tradename

EDQM

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C(CCS(=O)c2ccccc2)C(=O)N(N1c3ccccc3)c4ccccc4

InChI

1S/C23H20N2O3S/c26-22-21(16-17-29(28)20-14-8-3-9-15-20)23(27)25(19-12-6-2-7-13-19)24(22)18-10-4-1-5-11-18/h1-15,21H,16-17H2

InChI key

MBGGBVCUIVRRBF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfinpyrazone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuya Oguri et al.
Molecular cancer therapeutics, 7(5), 1150-1155 (2008-05-01)
We used the paclitaxel-resistant human small cell lung cancer subline PC-6/TAX1-1, selected from PC-6 cells by paclitaxel, to test whether MRP7/ABCC10 (ABCC10) confers paclitaxel resistance. We found that gene expression of both ABCB1/MDR1 (ABCB1) and ABCC10 was higher in PC-6/TAX1-1
Leslie R Harrold et al.
Arthritis research & therapy, 11(2), R46-R46 (2009-03-31)
Adherence to urate-lowering drugs (ULDs) has not been well evaluated among those with gout. Our aim was to assess the level and determinants of non-adherence with ULDs prescribed for gout. We identified persons using two integrated delivery systems aged 18
Rachida Tahar et al.
The American journal of tropical medicine and hygiene, 77(2), 221-227 (2007-08-11)
In vitro pyrimethamine response of Plasmodium falciparum isolates and dihydrofolate reductase (dhfr) gene sequences were analyzed in 2004-2005 and compared with our previous data. Most isolates (n = 103, all dhfr mutants) had 50% inhibitory concentrations (IC(50)s) > or =
Rina Daskalopoulos et al.
Glia, 39(2), 124-132 (2002-07-12)
Primary rat astrocyte cultures absorbed dehydroascorbic acid from the medium and reduced it to intracellular ascorbate. Uptake of dehydroascorbic acid (5-200 microM) was inhibited only partially by glucose (10 mM). The remaining glucose-insensitive component of dehydroascorbic acid uptake was inhibited
Shawn D Flanagan et al.
Pharmacology, 64(3), 126-134 (2002-02-09)
Recent studies in our laboratory have shown that the loop diuretic, furosemide, is actively secreted by Caco-2 cells and rat jejunal tissue. This active secretion could be the result of efflux transporters such as P-gp, MRP1 or MRP2 (cMOAT). To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service