Skip to Content
Merck
All Photos(2)

Key Documents

RDD043

Sigma-Aldrich

Acetylcholine chloride

≥99% (TLC), free-flowing, Redi-Dri

Synonym(s):

2-Acetoxyethyltrimethylammonium chloride, Trimethyl(2-acetoxyethyl)ammonium chloride, ACh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2CH2OCOCH3Cl-
CAS Number:
Molecular Weight:
181.66
Beilstein:
3571875
MDL number:
UNSPSC Code:
51151535
NACRES:
NA.25
Pricing and availability is not currently available.

product line

Redi-Dri

Quality Level

Assay

≥99% (TLC)

form

powder

quality

free-flowing

packaging

pkg of 100 g

color

white to faint yellow cast

mp

146-150 °C (lit.)

solubility

H2O: 100 mg/mL

density

1.27 g/cm3

storage temp.

room temp

SMILES string

[Cl-].CC(=O)OCC[N+](C)(C)C

InChI

1S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1

InChI key

JUGOREOARAHOCO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Acetylcholine Chloride, the chloride salt of acetylcholine, is a synthetic amino alcohol with cholinergic properties, making it a vital component in biochemical and neurotransmission studies. It replicates the effects of the endogenous acetylcholine compound, which plays a significant role in the immune system, inflammation, and allergy research. Acetylcholine chloride acts as a cholinergic neurotransmitter, binding to nicotinic and muscarinic acetylcholine receptors, facilitating the opening of calcium channels.Furthermore, Acetylcholine chloride (ACh chloride) functions as a potent cholinergic agonist, influencing the activity of dopaminergic (DAergic) neurons by stimulating nicotinic acetylcholine receptors (nAChRs). This feature is particularly relevant in studies related to the immune system and inflammation. Additionally, it demonstrates the capacity to inhibit the aggregation of p53 mutant peptide in vitro, which contributes to the exploration of biochemical and allergy research.

Application

Acetylcholine chloride has been used:
  • as an agent to trigger sudomotor axon reflex response during autonomic test[1]
  • to induce contraction amplitude of pyloric circular smooth muscle strip (PCSMS) in rats[2]
  • as a substrate for acetylcholinesterase[3]
  • to stimulate after discharge in Aplysia bag cell neurons[4]

Biochem/physiol Actions

Acetylcholine chloride, injected at 20 mg/kg body weight, reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis . The cholinergic anti-inflammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.
Endogenous neurotransmitter at cholinergic synapses; amplifies action potential of the sarcolemma thereby inducing muscle contractions.

Features and Benefits

  • Redi-Dri format for Improved Efficiency and Time Savings
  • Can be used in Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Caution

Very hygroscopic

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability of acetylcholine chloride solution in autonomic testing
Sletten DM, et al.
Journal of the Neurological Sciences, 234(1-2), 1-3 (2005)
Diarrhoea of famine and malnutrition--investigations using a rat model. 2--Ileal hypersecretion induced by starvation.
Young A and Levin RJ
Gut, 31(2), 162-169 (1990)
Acetylcholine Chloride: Physical Profile
Profiles of Drug Substances, Excipients, and Related Methodology, 31, 1-19 (2005)
Qing Cao et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 23(14), 1797-1805 (2016-12-04)
Radix Polygalae, the dried root of Polygala tenuifolia, has been extensively used as a traditional Chinese medicine for promoting intelligence and tranquilization. Polygalasaponins extracted from the root of P. tenuifolia possess evident anxiolytic and sedative-hypnotic activities. Previous studies have reported
Acetylcholine-evoked afterdischarge in Aplysia bag cell neurons
White SH and Magoski NS
American Journal of Physiology. Heart and Circulatory Physiology (2012)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service