Skip to Content
Merck
All Photos(1)

Documents

PHR3438

Supelco

Guanethidine Monosulfate

certified reference material, pharmaceutical secondary standard

Synonym(s):

Guanethidine monosulfate (1:1), 1-(2-Guanidinoethyl)octahydroazocine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H22N4 · H2SO4
CAS Number:
Molecular Weight:
296.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 181
traceable to USP 1301801

CofA

current certificate can be downloaded

packaging

pkg of 400 mg

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

OS(O)(=O)=O.NC(=N)NCCN1CCCCCCC1

InChI

1S/C10H22N4.H2O4S/c11-10(12)13-6-9-14-7-4-2-1-3-5-8-14;1-5(2,3)4/h1-9H2,(H4,11,12,13);(H2,1,2,3,4)

InChI key

YUFWAVFNITUSHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service