PHR2411
Guanosine
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Guanosine, 9-(β-D-Ribofuranosyl)guanine, Guanine-9-β-D-ribofuranoside
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
description
API family: adenosine
API family
adenosine
form
solid
CofA
current certificate can be downloaded
packaging
pkg of 100 mg
mp
250 °C (dec.) (lit.)
application(s)
pharmaceutical small molecule
storage temp.
-10 to -25°C
SMILES string
[H]O[H].NC1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1
InChI
1S/C10H13N5O5/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)/t3-,5-,6-,9-/m1/s1
InChI key
NYHBQMYGNKIUIF-UUOKFMHZSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatograms
application for LC-MSapplication for TLCapplication for HPLCapplication for HPLCShow MoreOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service