Skip to Content
Merck
All Photos(1)

Key Documents

PHR1648

Supelco

Omeprazole Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Omeprazole sulfone, 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulfonyl]-1H-benzimidazole, 5-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl}-1H-benzimidazole, Omeprazole sulphone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3O4S
CAS Number:
Molecular Weight:
361.42
Beilstein:
8347309
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. O0151000
traceable to USP 1478516

API family

omeprazole

CofA

current certificate can be downloaded

packaging

pkg of 30 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=S(C1=NC2=CC(OC)=CC=C2N1)(CC3=NC=C(C)C(OC)=C3C)=O

InChI

1S/C17H19N3O4S/c1-10-8-18-15(11(2)16(10)24-4)9-25(21,22)17-19-13-6-5-12(23-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)

InChI key

IXEQEYRTSRFZEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

It is an impurity of the substituted benzimidazole― omeprazole, that belongs to the class of proton pump inhibitors (PPIs). Omeprazole inhibits the secretion of gastric juice by inhibiting the activity of the enzyme H+/K+-ATPase (Hydrogen-Potassium Adenosine Triphosphates). Hence it is used widely for the treatment of diseases caused by excess secretion of gastric juices.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Analysis of omeprazole tablets for the separation and detection of omeprazole and its related impurities by reversed-phase high-performance liquid chromatographic (RP-HPLC) method[1]
  • Determination of omeprazole and its 11 related impurities in pharmaceutical formulations by RP-HPLC method based on quality-by-design approach (QbD)
  • Development of an ultra-high performance liquid chromatography-time of flight-mass spectrometry (UHPLC-TOF-MS) based method to estimate omeprazole and its related impurities in pharmaceutical formulations
  • Separation and detection of omeprazole and its metabolites in human urine samples by capillary zone electrophoresis (CZE) combined with UV-Visible detector
  • Estimation of related impurities of omeprazole and domperidone in their combined dosage form using the RP-HPLC method

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3168 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of a Precise single HPLC Method for Determination of Omeprazole and its related compound in pharmaceutical formulation
Trivedi HK and Patel MC
International Journal of ChemTech Research, 2, 1355-1367 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service