Skip to Content
Merck
All Photos(1)

Key Documents

PHR1384

Supelco

Amitriptyline hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Amitriptyline hydrochloride, 10,11-Dihydro-N,N-dimethyl-5H-dibenzo[a,d]cycloheptane-Δ5γ-propylamine hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-1-propanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N · HCl
CAS Number:
Molecular Weight:
313.86
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 16
traceable to Ph. Eur. Y0000563
traceable to USP 1029002

API family

amitriptyline

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].CN(C)CC\C=C1\c2ccccc2CCc3ccccc13

InChI

1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H

InChI key

KFYRPLNVJVHZGT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Amitriptyline hydrochloride is a drug, and a tricyclic antidepressant with anticholinergic activity.[1] It is widely used in combination with chlordiazepoxide 7-chloro-2-(methyl-amino)-5-phenyl-3H-1,4-benzodiazepine-4-oxide for the treatment of patients with moderate to severe depression associated with moderate to severe anxiety.[2]

Application

Amitriptyline hydrochloride may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique and reversed-phase high-performance liquid chromatography.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Tricyclic antidepressant; inhibits the norepinephrine and serotonin transporters with Kis of 100 nM and 14.7 nM, respectively; high in vitro affinity for α1-adrenoceptors, serotonin and muscarinic acetylcholine receptors.
Tricyclic antidepressant; norepinephrine and serotonin transporter inhibitor; high in vitro affinity for α1-adrenoceptors, serotonin and muscarinic acetylcholine receptors.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1640 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric and chromatographic simultaneous estimation of amitriptyline hydrochloride and chlordiazepoxide in tablet dosage forms
Patel S, et al.
Indian Journal of Pharmaceutical Sciences, 71(4), 472-472 (2009)
Simultaneous spectrophotometric determination of amitriptyline hydrochloride and chlordiazepoxide in pharmaceutical tablets by multivariate calibration method
Sarrafi MHA, et al.
Journal of Chemistry, 6(S1), S111-S116 (2009)
Katja Kovacic et al.
The Journal of pediatrics, 164(5), 1104-1109 (2014-03-13)
To compare children with primary, chronic idiopathic nausea to those with secondary nausea associated with functional abdominal pain. Retrospective chart review of 45 children with a primary complaint of chronic nausea several times per week. Comparisons were made to prospectively
Mariola Herbet et al.
Acta poloniae pharmaceutica, 70(5), 913-917 (2013-10-24)
The aim of the present study was to assess the impact of combined therapy with rosuvastatin (10 mg/kg) and amitriptyline (10 mg/kg) on oxidation-reduction status in the blood of rats. After 2-week application of drugs alone or their combination, the
Grzegorz Jagla et al.
Pharmacological reports : PR, 66(3), 459-465 (2014-06-07)
The therapy of neuropathic pain may include the use of co-analgesics, such as antidepressants, however, their desired analgesic effect is associated with significant side effects. An alternative approach to this is their local administration which has been proposed, but there

Articles

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service