PHR1358
DMDM Hydantoin
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
DMDMH
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
API family
dmdm hydantoin
CofA
current certificate can be downloaded
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-30°C
General description
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards..
Application
DMDM Hydantoin may be used as a pharmaceutical reference standard for the determination of the analyte in cosmetics by thermal desorption electrospray ionization mass spectrometry (TD-ESI-MS).
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Analysis Note
These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Footnote
To see an example of a Certificate of Analysis for this material enter LRAA1411 in the slot below. This is an example certificate only and may not be the lot that you receive.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Using thermal desorption electrospray ionization mass spectrometry to rapidly determine antimicrobial preservatives in cosmetics
Rapid Communications in Mass Spectrometry, 30(21), 2315-2322 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service