Skip to Content
Merck
All Photos(1)

Key Documents

PHR1332

Supelco

L-Proline

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

L-Proline, (S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80810
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. P3350000
traceable to USP 1568506

API family

proline

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

228 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Proline is a non-essential amino acid. Peptides bond to proline, making it a useful building block for proteins. It can be used as a cell culture media component for the commercial biomanufacturing of therapeutic recombinant proteins and monoclonal antibodies.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

L-Proline may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3756 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proline
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 3683-3683 (2018)
Daniel R Knight et al.
Applied and environmental microbiology, 81(1), 119-123 (2014-10-19)
Clostridium difficile is an important enteric pathogen of humans and the cause of diarrhea and enteritis in neonatal pigs. Outside Australia, prevalence in piglets can be up to 73%, with a single PCR ribotype (RT), 078, predominating. We investigated the
Mattias Mandorfer et al.
AIDS patient care and STDs, 28(5), 221-227 (2014-05-07)
The aim of this study was to assess the therapeutic potential of telaprevir (TPV)/boceprevir (BOC)-based triple-therapy in a complete cohort of HIV/chronic hepatitis C co-infected patients (HIV/HCV). Moreover, a case series of four HIV/HCV genotype (HCV-GT)1 patients with rapid virologic
John M Curto et al.
Organic letters, 16(7), 1948-1951 (2014-03-29)
The first asymmetric synthesis of α-allyl-α-aryl α-amino acids by means of a three-component coupling of α-iminoesters, Grignard reagents, and cinnamyl acetate is reported. Notably, the enolate from the tandem process provides a much higher level of reactivity and selectivity than
Marco Scocchi et al.
Cellular and molecular life sciences : CMLS, 68(13), 2317-2330 (2011-05-20)
Proline-rich antimicrobial peptides are a group of cationic host defense peptides of vertebrates and invertebrates characterized by a high content of proline residues, often associated with arginine residues in repeated motifs. Those isolated from some mammalian and insect species, although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service