Skip to Content
Merck
All Photos(1)

Documents

PHR1293

Supelco

Nimodipine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Nimodipine, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-methoxyethyl 1-methylethyl ester, Isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-(m-nitrophenyl)-3,5-pyridinedicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O7
CAS Number:
Molecular Weight:
418.44
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 651
traceable to Ph. Eur. N0850000
traceable to USP 1463858

API family

nimodipine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COCCOC(=O)C1=C(C)NC(C)=C(C1c2cccc(c2)[N+]([O-])=O)C(=O)OC(C)C

InChI

1S/C21H26N2O7/c1-12(2)30-21(25)18-14(4)22-13(3)17(20(24)29-10-9-28-5)19(18)15-7-6-8-16(11-15)23(26)27/h6-8,11-12,19,22H,9-10H2,1-5H3

InChI key

UIAGMCDKSXEBJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nimodipine is a well-known calcium antagonist and is a moderate cerebral vasodilator, in addition to being an inhibitor of cerebrovascular reactivity. It is suggested to be cytoprotective by causing a decrease in the influx of calcium into the nerve cells. It is accepted for use in the management of delayed ischaemic dysfunction and is commonlly used in the studies of brain function impairment and dementia in old age.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Nimodipine can be used as a pharmaceutical reference standard in the determination of the analyte in pharmaceutical formulations and combined dosage forms by various electrochemical and spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0583 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile spectrophotometric determination of nimodipine and nitrazepam in pharmaceutical preparations
Revanasiddappa HD, et al.
Analele Universitatii Bucuresti: Chimie , 20(1), 189-196 (2011)
The atomic absorption spectrophotometric method for indirect determination of nimodipine in tablets
Canlica M and Islimyeli S
Turkish Journal of Chemistry, 29(2), 141-146 (2005)
Nitrogen-doped graphene modified electrode for nimodipine sensing
Lei W, et al.
Sensors and Actuators B, Chemical, 212(1), 207-213 (2015)
Assay of nimodipine-an anti hypertensive drug, in bulk form and pharmaceutical formulations by cathodic adsorptive stripping voltammetry
Gupta VK, et al.
International Journal of Electrochemical Science, 6(1), 37-51 (2011)
Effect of oral nimodipine on cerebral infarction and outcome after subarachnoid haemorrhage: British aneurysm nimodipine trial.
Pickard JD, et al.
BMJ (Clinical Research ed.), 298(6674), 636-642 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service