Skip to Content
Merck
All Photos(1)

Documents

PHR1228

Supelco

Dexpanthenol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-Panthenol, (R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, D-Pantothenyl alcohol, Dexpanthenol, Provitamin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
Beilstein:
1724947
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. D0730000
traceable to USP 1179504

API family

dexpanthenol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

1.20 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-30°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCCO

InChI

1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/t7-/m0/s1

InChI key

SNPLKNRPJHDVJA-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dexpanthenol is an alcohol analog of pantothenic acid and an important component of parenterals, local cosmetic preparations and multi-vitamins. It has as much biological activity as pantothenic acid and is found to be stable in aqueous solution.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Dexpanthenol may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0292 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of dexpanthenol, lidocaine hydrochloride, and mepyramine maleate in combined pharmaceutical gel by capillary electrophoresis
Akyil BG, et al.
Turkish Journal of Chemistry, 38(5), 756-764 (2014)
Development and validation of reversed phase high performance liquid chromatography method for determination of dexpanthenol in pharmaceutical formulations.
Kulikov U A and Zinchenko A A.
Journal of Pharmaceutical and Biomedical Analysis, 43(3), 983-988 (2007)
Determination of eight water-and fat-soluble vitamins in multi-vitamin pharmaceutical formulations by high-performance liquid chromatography
Moreno P and Salvado V
Journal of Chromatography A, 870(1-2), 207-215 (2000)
M Göbbels et al.
Klinische Monatsblatter fur Augenheilkunde, 209(2-3), 84-88 (1996-08-01)
In this controlled, randomized, double-masked study the effect of dexpanthenol-containing artificial tears (Siccaprotect) on patients with dry eyes was examined. 50 patients applied either dexpanthenol-containing artificial tears (Siccaprotect) or the identical, but free of dexpanthenol, eye drops five times daily
M Sachs et al.
Zeitschrift fur Ernahrungswissenschaft, 29(4), 270-283 (1990-12-01)
The aim of this study was the examination of the metabolism and mechanism of action of D-pantothenyl alcohol in patients with postoperative intestinal atony. Seven metabolically healthy patients were examined on the 4th day following colorectal surgery, before bowel activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service