Skip to Content
Merck
All Photos(1)

Key Documents

PHR1181

Supelco

2-Methylimidazole (Ondansetron Impurity F)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Methylimidazole, Ondansetron impurity F, 2-Methylglyoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein:
1368
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001320

vapor pressure

<1 mmHg ( 0 °C)

API family

ondansetron

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

267-268 °C (lit.)

mp

142-143 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cc1ncc[nH]1

InChI

1S/C4H6N2/c1-4-5-2-3-6-4/h2-3H,1H3,(H,5,6)

InChI key

LXBGSDVWAMZHDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9000 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jerry D Johnson et al.
Journal of toxicology and environmental health. Part A, 65(12), 869-879 (2002-06-25)
The toxicokinetics of 2-methylimidazole (2-MI) were studied in male and female Fischer 344 rats after a single iv dose of 10 mg/kg or gavage dose of 25, 50, or 100 mg/kg. The 2-MI was formulated in 0.05 M phosphate-buffered saline
P C Chan et al.
Archives of toxicology, 82(6), 399-412 (2007-10-10)
2-Methylimidazole (2MI) has been identified as a by-product of fermentation and is detected in foods and mainstream and side-stream tobacco smoke. It is used in the manufacture of pharmaceuticals, photographic chemicals, dyes and pigments, agricultural chemicals, and rubber. Carcinogenicity studies
M van Gastel et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 4(3), 257-265 (1999-08-10)
Frozen solutions of the azurin mutant His117Gly in the presence of excess of methyl-substituted imidazoles have been investigated by electron spin-echo envelope modulation (ESEEM) spectroscopy at 9 GHz. The addition of imidazole is known to reconstitute a blue-copper site and
Kiriaki Pegklidou et al.
Journal of agricultural and food chemistry, 56(12), 4482-4490 (2008-05-23)
The potential role of 2-methyl imidazole in improving lycopene production by Blakeslea trispora with regards to yield, selectivity, and safety aspects was investigated in batch culture. Optimization of the bioprocess conditions in terms of (a) (+) and (-) strain ratio
Jianfeng Li et al.
Inorganic chemistry, 49(5), 2398-2406 (2010-01-29)
The X-ray characterization of the five-coordinate picket-fence porphyrin complex, [Co(TpivPP)(2-MeHIm)], is reported. The complex has the displacement of cobalt from the porphyrin plane = 0.15 A, and Co-N(Im) = 2.145(3) and (Co-N(p))(av) = 1.979(3) A. This five-coordinate complex, in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service