Skip to Content
Merck
All Photos(1)

Key Documents

PHR1166

Supelco

Cetyl Palmitate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Palmityl palmitate, Cetyl palmitate, Hexadecyl hexadecanoate, Palmitic acid palmityl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2(CH2)15CH3
CAS Number:
Molecular Weight:
480.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000074
traceable to USP 1103105

API family

cetyl palmitate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

55-56 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

InChI

1S/C32H64O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-34-32(33)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3

InChI key

PXDJXZJSCPSGGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cetyl Palmitate is an ester of palmitic acid, obtained via the reaction of cetyl alcohol and palmitic acid. It is an ingredient of many cosmetic preparations.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7179 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Fiber Finish Technology (1997)
A Consumerys Dictionary of Household, Yard and Office Chemicals: Complete Information About Harmful and Desirable Chemicals Found in Everyday Home Products, Yard Poisons, and Office Polluters (2007)
Paolo Blasi et al.
International journal of pharmaceutics, 419(1-2), 287-295 (2011-08-11)
The aim of this study was to optimize the formulation of lipid nanoparticles (NPs), intended for brain targeting, with the aid of a computer generated experimental design. The high pressure homogenization technique, selected for this purpose, was suitable to formulate
G Lukowski et al.
International journal of pharmaceutics, 196(2), 201-205 (2000-03-04)
Solid lipid nanoparticles (SLN) as alternative intravenous colloidal drug carriers were produced by high pressure homogenisation of the melted lipid cetylpalmitate. The crystallographic properties of the used cetylpalmitate SLN were characterised by small angle X-ray scattering (SAXS) and X-ray diffraction
C Carbone et al.
European journal of medicinal chemistry, 49, 110-117 (2012-01-17)
The objective of this study was the preparation, physico-chemical characterization and statistical optimization of cationic solid lipid nanoparticles (SLN) prepared by the PIT method as potential carrier for gene therapy, emphasizing the application of factorial design in such a kind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service