Skip to Content
Merck
All Photos(1)

Key Documents

PHR1053

Supelco

Albuterol sulfate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Albuterol Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44N2O10S
CAS Number:
Molecular Weight:
576.70
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 302
traceable to Ph. Eur. S0150000
traceable to USP 1012633

API family

albuterol, salbutamol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)O.N(C(C)(C)C)CC(O)c2cc(c(cc2)O)CO.N(C(C)(C)C)CC(O)c1cc(c(cc1)O)CO

InChI

1S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17H,7-8H2,1-3H3;(H2,1,2,3,4)

InChI key

BNPSSFBOAGDEEL-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

General description

Albuterol sulfate is a β2 adrenergic agonist and a bronchodilating agent, which is used in the treatment of asthma.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Albuterol sulfate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by ultra-performance liquid chromatographic method with ultraviolet detection (UPLC-UV).
Albuterol sulfate may be used as a reference standard in determining the concentration of albuterol sulfate present in inhalers using reversed-phase high performance liquid chromatography (RP-HPLC).
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7128 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of albuterol sulfate and its related substances in albuterol sulfate inhalation solution, 0.5% by RP-HPLC
Erram.VS, et al.
Journal of Pharmaceutical and Biomedical Analysis, 40(4), 864-874 (2006)
Supercritical and near-critical carbon dioxide assisted low-temperature bubble drying
Sievers.ER, et al.
Industrial & Engineering Chemistry Research, 39(12), 4831-4836 (2000)
Comparison of conventional HPLC with UPLC method for determination of albuterol sulfate and it?s impurities in pharmaceutical formulation.
Dalvi PN, et al.
Orient. J. Chem., 24(2), 537-544 (2008)
B Padhukasahasram et al.
The pharmacogenomics journal, 14(4), 365-371 (2014-01-15)
Inhaled short-acting beta-agonist (SABA) medication is commonly used in asthma patients to rapidly reverse airway obstruction and improve acute symptoms. We performed a genome-wide association study of SABA medication response using gene-based association tests. A linear mixed model approach was
Huan Wang et al.
Biosensors & bioelectronics, 49, 14-19 (2013-05-28)
A multiplexed electrochemical biosensor has been developed for fast and sensitive detection of ractopamine (RAC), salbutamol (SAL) and clenbuterol (CLB) based on reduced graphene oxide (rGO) and silver-palladium alloy nanoparticles (AgPd NPs). In this paper, rGO with high conductivity was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service