Skip to Content
Merck
All Photos(1)

Key Documents

PHR1042

Supelco

Prednisone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1,4-Pregnadiene-17α,21-diol-3,11,20-trione, 1-Cortisone, 17α,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Dehydrocortisone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
Beilstein:
2065301
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 553
traceable to Ph. Eur. P2900000
traceable to USP 1559006

API family

prednisone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

236-238 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]([C@@](CC[C@@]3(C(CO)=O)O)([H])[C@]3(C)C4)([H])[C@]2([H])C4=O)=C1

InChI

1S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,18,22,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1

InChI key

XOFYZVNMUHMLCC-ZPOLXVRWSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Prednisone may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by spectrophotometry and chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8969 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald de Wit
European journal of cancer (Oxford, England : 1990), 41(4), 502-507 (2005-03-02)
Until now, the use of systemic chemotherapy for advanced androgen-independent prostate cancer has had very little to offer to patients. However, in 2004, two large randomised trials investigating docetaxel vs. mitoxantrone have both demonstrated survival improvements, and, in one of
O Teuffel et al.
Leukemia, 25(8), 1232-1238 (2011-04-30)
This systematic review and meta-analysis compared the efficacy and toxicity of dexamethasone (DEX) versus prednisone (PRED) for induction therapy in childhood acute lymphoblastic leukemia (ALL). We searched biomedical literature databases and conference proceedings for randomized controlled trials comparing DEX and
Jörg D Leuppi et al.
JAMA, 309(21), 2223-2231 (2013-05-23)
International guidelines advocate a 7- to 14-day course of systemic glucocorticoid therapy in acute exacerbations of chronic obstructive pulmonary disease (COPD). However, the optimal dose and duration are unknown. To investigate whether a short-term (5 days) systemic glucocorticoid treatment in
Ian F Tannock et al.
The Lancet. Oncology, 14(8), 760-768 (2013-06-08)
Docetaxel plus prednisone is standard first-line chemotherapy for men with metastatic castrate-resistant prostate cancer. Aflibercept is a recombinant human fusion protein that binds A and B isoforms of VEGF and placental growth factor, thereby inhibiting angiogenesis. We assessed whether the
Charles J Ryan et al.
The Lancet. Oncology, 16(2), 152-160 (2015-01-21)
Abiraterone acetate plus prednisone significantly improved radiographic progression-free survival compared with placebo plus prednisone in men with chemotherapy-naive castration-resistant prostate cancer at the interim analyses of the COU-AA-302 trial. Here, we present the prespecified final analysis of the trial, assessing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service