Skip to Content
Merck
All Photos(1)

Key Documents

PHR1028

Supelco

1,4-Benzoquinone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

p-Benzoquinone, Quinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(=O)2
CAS Number:
Molecular Weight:
108.09
Beilstein:
773967
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1056504

vapor density

3.73 (vs air)

vapor pressure

0.1 mmHg ( 25 °C)

CofA

current certificate can be downloaded

autoignition temp.

815 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

113-115 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C=CC(=O)C=C1

InChI

1S/C6H4O2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

AZQWKYJCGOJGHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Benzoquinone is a cyclic conjugated diketone. It belongs to the class of quinones, naturally occurring in plants, fungi, and bacteria.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7811 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quinone chemistry and toxicity
Monks JT, et al.
Toxicology and Applied Pharmacology, 112(1), 2-16 (1992)
Takafumi Hasegawa et al.
Journal of neurochemistry, 87(2), 470-475 (2003-09-27)
Oxidized metabolites of dopamine, known as dopamine quinone derivatives, are thought to play a pivotal role in the degeneration of dopaminergic neurons. Although such quinone derivatives are usually produced via the autoxidation of catecholamines, tyrosinase, which is a key enzyme
Vandana Bhalla et al.
Dalton transactions (Cambridge, England : 2003), 42(13), 4464-4469 (2013-02-16)
The binding behavior of nanoaggregates of pentacenequinone derivative 1 having thiophene groups has been investigated toward various cations (Fe(3+), Fe(2+), Pd(2+), Mg(2+), Cd(2+), Hg(2+), Ni(2+), Zn(2+), Cu(2+), Pb(2+), Co(2+), K(+), Na(+) and Li(+)) using UV-vis and fluorescence spectroscopy. Among the
K He et al.
Biochemical pharmacology, 62(2), 191-198 (2001-06-08)
Oxidative ring opening of troglitazone (TGZ)(1) a thiazolidine 2,4-dione derivative used for the treatment of type II diabetes mellitus, leads to the formation of a quinone metabolite. The formation of TGZ quinone was shown to be NADPH dependent and to
S Saif Hasan et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(11), 4297-4302 (2013-02-27)
As much as two-thirds of the proton gradient used for transmembrane free energy storage in oxygenic photosynthesis is generated by the cytochrome b6f complex. The proton uptake pathway from the electrochemically negative (n) aqueous phase to the n-side quinone binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service