Skip to Content
Merck
All Photos(2)

Key Documents

P6863

Sigma-Aldrich

PDAM

≥70% purity (HPLC), for HPLC derivatization, solid

Synonym(s):

1-Pyrenyldiazomethane, Pyrene, 1-(diazomethyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H10N2
CAS Number:
Molecular Weight:
242.27
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

Product Name

PDAM, for HPLC derivatization

grade

for HPLC derivatization

Quality Level

Assay

≥70% (HPLC)

form

solid

technique(s)

titration: suitable

λmax

338-344 nm

fluorescence

λem 371-379 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

dry ice

storage temp.

−20°C

SMILES string

[N-]=[N+]=Cc1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C17H10N2/c18-19-10-14-7-6-13-5-4-11-2-1-3-12-8-9-15(14)17(13)16(11)12/h1-10H

InChI key

PEIBAWRLFPGPAT-UHFFFAOYSA-N

Application

Fluorescent diazoalkane useful as an HPLC derivatization reagent for carboxylic acid detection with better detection limit parameters and greater chemical stability than the commonly used 9-anthryldiazomethane.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Iohan et al.
Journal of chromatography, 564(1), 27-41 (1991-03-08)
A new procedure is described for the detection of the acidic metabolites of cortisol (cortoic acids) as the pyrenylmethyl-21-oic esters. The derivatizing reagent, diazomethylpyrene, was prepared by an improved procedure. The reagent was used at room temperature, required no catalyst
Carlos García et al.
Biological research, 36(2), 171-183 (2003-09-30)
Diarrhetic shellfish poisoning (DSP) is a gastrointestinal disease caused by lipid soluble polyether toxins produced by dinoflagellates and accumulated in shellfish. Diarrhetic shellfish poisoning is a worldwide threat to public health and the shellfish industry. To date, only four lipid
Liquid chromatographic determination of biotin by using 1-pyrenyldiazomethane as a pre-column fluorescent labelling reagent.
T Yoshida et al.
Journal of chromatography, 456(2), 421-426 (1988-12-16)
O L Brekke et al.
Journal of lipid research, 38(9), 1913-1922 (1997-11-05)
A highly sensitive method to determine agonist-induced release of endogenous fatty acids from cells in culture was developed using high-performance liquid chromatography and fluorescence detection. Fatty acids were selectively derivatized with 1-pyrenyldiazomethane and separated on a LC18 reversed phase column
Junko Amano et al.
Analytical chemistry, 82(20), 8738-8743 (2010-09-25)
Glycoproteomics holds the promise of new advances in medical technology. However, mass spectrometry has limitations for the structural determination of glycosylated peptides because the hydrophilic nature of the oligosaccharide moiety in glycopeptides is disadvantageous for ionization, and glycopeptides ionize much

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service