Skip to Content
Merck
All Photos(4)

Key Documents

P5504

Sigma-Aldrich

Potassium phosphate dibasic trihydrate

Potassium phosphate dibasic trihydrate

ReagentPlus®, ≥99.0%

Synonym(s):

Dipotassium hydrogen phosphate trihydrate, Dipotassium phosphate, di-Potassium hydrogen phosphate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4 · 3H2O
CAS Number:
Molecular Weight:
228.22
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

pH

9.2-9.4 (25 °C, 50 g/L)

solubility

water: 100 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

O.O.O.[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P.3H2O/c;;1-5(2,3)4;;;/h;;(H3,1,2,3,4);3*1H2/q2*+1;;;;/p-2

InChI key

XQGPKZUNMMFTAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phosphate solution have very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography. Potassium phosphate dibasic trihydrate is a key component of potassium phosphate buffer, phosphate-buffered saline (PBS), and phosphate-buffered EDTA (PBE). It is extensively used in the preparation of biological samples for various biological experiments.[1] Potassium phosphate buffer is also used in methods such as equilibrium dialysis, high-performance liquid chromatography (HPLC), and affinity capillary electrophoresis (ACE) to study the characteristics of drug-protein interactions in the body.[2] Potassium phosphate buffer solutions consist of mixtures of the potassium phosphate monobasic and potassium phosphate dibasic forms.[3]

Application

Potassium phosphate dibasic trihydrate has been used in the preparation of potassium phosphate buffer for the resuspension of polyelectrolyte capsules.[4] It has also been used as a buffer during glutaminase‑1 (GLS-1) enzyme assay and GLS-2 enzyme assay.[5]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction between Potassium Phosphate Buffer Solution and Modeling Cell Membrane Investigated by Sum Frequency Generation Vibrational Spectroscopy
Weilai Y, et al.
Chinese Journal of Chemical Physics, 28(4), 518-524 (2015)
Daniel Valdepérez et al.
Journal of colloid and interface science, 474, 1-8 (2016-04-19)
Polyelectrolyte hollow capsules are versatile platforms typically used for encapsulation of a wide variety of macromolecules in their cavity. The polymer shell of these capsules as composed by alternating layers of oppositely charged polyelectrolytes also allows for adding additional functionalities.
Antoine Desrut et al.
Journal of experimental botany, 71(22), 7301-7315 (2020-08-30)
Plants live in close relationships with complex populations of microorganisms, including rhizobacterial species commonly referred to as plant growth-promoting rhizobacteria (PGPR). PGPR are able to improve plant productivity, but the molecular mechanisms involved in this process remain largely unknown. Using
Imidazolium-based Ionic Liquids as Adjuvants to Form Polyethylene Glycol with Salt Buffer Aqueous Biphasic Systems
Santos JHPM, et al.
Journal of Chemical and Engineering Data, 65(8), 3794?3801-3794?3801 (2020)
Michael J Soth et al.
Journal of medicinal chemistry, 63(21), 12957-12977 (2020-10-30)
Inhibition of glutaminase-1 (GLS-1) hampers the proliferation of tumor cells reliant on glutamine. Known glutaminase inhibitors have potential limitations, and in vivo exposures are potentially limited due to poor physicochemical properties. We initiated a GLS-1 inhibitor discovery program focused on

Protocols

Enzymatic Assay of Superoxide Dismutase

Related Content

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service