Skip to Content
Merck
All Photos(1)

Key Documents

NIST8375

Microbial Genomic DNA Standards for Sequencing Performance Assessment

(MG-001, MG-002, MG-003, MG-004)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

manufacturer/tradename

NIST®

technique(s)

electrophoresis: suitable

application(s)

genomic analysis

format

multi-component solution

General description

This microbial genomic DNA standard is a reference material (RM) that consists of four vials. The genomic DNA is extracted from cell cultures. Each vial contains approximately 2 µg of microbial genomic DNA in TE buffer (10 mM TRIS, 1 mM EDTA, pH 8.0).

  • MG-001 Salmonella Typhimurium LT2
  • MG-002 Staphylococcus aureus
  • MG-003 Pseudomonas aeruginosa
  • MG-004 Clostridium sporogenes
NIST8375_Cert

NIST8375_SDS

Application

This reference material (RM) is suitable for validation, optimization, process evaluation, and performance assessment of microbial whole genome sequencing.

Features and Benefits

  • Enables the assessment of the performance of high-throughput microbial DNA sequencing methods.
  • It does not assess pre-analytical steps such as DNA extraction as the RM is extracted DNA.
  • Challenges sequencing library preparation, sequencing machines, base calling algorithms, and subsequent bioinformatics analyses.
  • This genomic DNA is extracted from cell cultures.

Other Notes

  • Refer to the SDS for additional information on biomaterials, disposal, and transport.
  • Details on expiration, maintenance, usage, storage, preparation, stability, homogeneity, and purity are available in the certificate.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service