Skip to Content
Merck
All Photos(1)

Key Documents

NIST1886B

White Portland Cement

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 20 g

manufacturer/tradename

NIST®

format

matrix material

storage temp.

15-25°C

General description

Other Notes

Certified for the analytes listed below.
Silicon dioxide (SiO2), Aluminium di oxide (Al2O3), Ferric oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sodium oxide (Na2O), Potassium oxide (K2O), Titanium di oxide (TiO2), Phosphorus pentoxide (P2O5), Manganese(III) oxide (Mn2O3), Chromium(III) oxide (Cr2O3), Strontium oxide (SrO), Chloride (Cl)

Reference/Informational value is provided for the following analytes.
Zinc oxide (ZnO), Barium oxide (BaO), Sulfide sulfur (S2-), Calcium oxide (CaO), Fluoride (F), Mercury (Hg)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service