Skip to Content
Merck
All Photos(1)

Key Documents

NIST1400

Bone ash

NIST® SRM® 1400

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

shelf life

5 yr

packaging

pkg of 50 g

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

application(s)

forensics and toxicology

format

matrix material

General description

This bone ash standard reference material (SRM) consists of ash that was blended to a high degree of homogeneity and bottled in 50g units.

NIST1400_Cert

NIST1400_SDS

Application

This bone ash standard reference material (SRM) is intended primarily for use in evaluating analytical methods for the determination of selected trace elements in bone and in the material of a similar matrix.

Features and Benefits

  • The certified values for this material are given in the NIST Certificate of Analysis.

Other Notes

  • Details on expiry, usage, and dissolution procedure are available in the NIST certificate.
  • Notes on handling, storage, toxicological information, regulatory, transportation, and disposal are provided in the SDS.
Certified for the analytes listed below.
Calcium (Ca), Magnesium (Mg), Phosphorus (P), Iron (Fe), Lead (Pb), Potassium (K), Strontium (Sr), Zinc (Zn)

Reference/Informational value is provided for the following analytes.
Silicon (Si), Sodium (Na), Aluminum (Al), Arsenic (As), Cadmium (Cd), Copper (Cu), Flurine (F), Manganese (Mn), Selenium (Se)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of neutron activation analysis to bone samples
Zamboni C B, et al.
Journal of radioanalytical and nuclear chemistry, 269, 331?334- 331?334 (2006)
Determination of toxic and other trace elements in calcium-rich materials using cloud point extraction and inductively coupled plasma emission spectrometry.
Borkowska-Burnecka J, et al.
Journal of Hazardous Materials, 182(1-3), 477-483 (2010)
Chemical compositional changes in archaeological human bones due to diagenesis: Type of bone vs soil environment
Olalla L, et al.
The Journal of Arthroplasty, 67, 43-51 (2016)
Comparison of matrix effects on portable and stationary XRF spectrometers for cultural heritage samples
Pessanha S, et al.
Applied Microbiology, 497?505-497?505 (2009)
Determination of trace elements and calcium in bone of the human iliac crest by atomic absorption spectrometry.
Scancar J, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 293(1-2), 187-197 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service