N44
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified
viscosity 71.11 mPa.s (25 °C)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
form
liquid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
viscosity
71.11 mPa.s(25 °C)
Looking for similar products? Visit Product Comparison Guide
General description
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yanfeng Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 65, 104778-104778 (2020-01-22)
The need of in vitro alternative methods has been increasing in toxicology research as well as in cosmetic industry in China recently. Following the establishment of China EpiSkin™ skin corrosion and irritation testing methods, both as stand-alone in vitro tests
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service