Skip to Content
Merck
All Photos(1)

Key Documents

N0590200

Nicotine ditartrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Nicotine hydrogen tartrate salt, (−)-1-Methyl-2-(3-pyridyl)pyrrolidine (+)-bitartrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2 · 2C4H6O6
CAS Number:
Molecular Weight:
462.41
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nicotine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1CCC[C@H]1C2=CC=CN=C2.C

InChI

1S/C10H14N2.CH4/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;/h2,4,6,8,10H,3,5,7H2,1H3;1H4/t10-;/m0./s1

InChI key

RYYKFQGHMYGLEL-PPHPATTJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicotine ditartrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Prototype nicotinic acetylcholine receptor agonist; naturally occurring isomer.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

289.8 °F

Flash Point(C)

143.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueliang Shang et al.
Molecular neurobiology, 54(6), 4644-4658 (2016-07-14)
The aim of this study was to examine if nicotine was able to improve cognition deficits in a mouse model of chronic mild stress. Twenty-four male C57BL/6 mice were divided into three groups: control, stress, and stress with nicotine treatment.
Gabriel Quiroz et al.
Frontiers in pharmacology, 10, 1429-1429 (2019-12-19)
Brain nicotinic acetylcholine receptors (nAChRs), a heterogeneous family of pentameric acetylcholine-gated cation channels, have been suggested as molecular targets for the treatment of alcohol abuse and dependence. Here, we examined the effect of the competitive nAChR antagonist UFR2709 on the
José-Rubén García-Montes et al.
Molecular neurobiology, 56(6), 4037-4050 (2018-09-28)
L-DOPA is the main pharmacological therapy for Parkinson's disease. However, long-term exposure to L-DOPA induces involuntary movements termed dyskinesia. Clinical trials show that dyskinesia is attenuated by metabotropic glutamate receptor type 5 (mGluR5) antagonists. Further, the onset of dyskinesia is
Marianne Husson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(17), 3465-3477 (2020-03-19)
Nicotine addiction, through smoking, is the principal cause of preventable mortality worldwide. Human genome-wide association studies have linked polymorphisms in the CHRNA5-CHRNA3-CHRNB4 gene cluster, coding for the α5, α3, and β4 nicotinic acetylcholine receptor (nAChR) subunits, to nicotine addiction. β4*nAChRs
Monica M Arnold et al.
Neuropharmacology, 85, 293-304 (2014-06-08)
Tobacco use is the leading cause of preventable death. Although the health risks are well known, cessation rates remain low. Whereas behavioral and neuroanatomical studies on tobacco addiction conventionally use nicotine, there is evidence that other constituents, such as monoamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service