Skip to Content
Merck
All Photos(2)

Key Documents

M2649

Sigma-Aldrich

Melanin from Sepia officinalis

99% purity (TLC), powder

Synonym(s):

Melanin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Melanin from Sepia officinalis, 99% (TLC)

Quality Level

Assay

99% (TLC)

form

powder

solubility

hydrogen peroxide: 1 mg/mL, dark brown

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[nH]1c2c3c(c4c5c([nH]c4)c([c]([c](c5c3c1)=O)=O)C)[c]([c](c2C)=O)=O

InChI

1S/C18H10N2O4/c1-5-13-9-7(3-19-13)12-10-8(11(9)17(23)15(5)21)4-20-14(10)6(2)16(22)18(12)24/h3-4,19-20H,1-2H3

InChI key

XUMBMVFBXHLACL-UHFFFAOYSA-N

General description

Melanin is a ubiquitous and prominent pigment responsible for color of skin, hair and eyes. Melanin is classified into eumelanin and pheomelanin. Eumelanin is black to dark brown pigment and is present in black hair and retina of the eye. Pheomelanin is yellow to reddish brown pigment present is red hair and red feathers. Melanins are high molecular weight pigments synthesized from phenolic or indolic precursors and is widely present in animals, plants and microorganisms.

Application

Melanin from Sepia officinalis has been used:
  • as a natural pigment for Raman spectroscopy measurements
  • in competitive enzyme linked immunosorbent assay (ELISA) for the characterization of phage antibodies
  • as an external standard for spectrophotometrically determining the melanin production in B16F10 cells

Biochem/physiol Actions

Melanin scavenges active chemical species and acts as a sunscreen.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bor-Kai Hsiung et al.
The Journal of experimental biology, 218(Pt 22), 3632-3635 (2015-10-10)
Melanin pigments are broadly distributed in nature - from bacteria to fungi to plants and animals. However, many previous attempts to identify melanins in spiders were unsuccessful, suggesting that these otherwise ubiquitous pigments were lost during spider evolution. Yet, spiders
Optical properties of melanin in the skin and skinlike phantoms.
Bashkatov AN
SPIE Proc. (2000)
Influence of rosmarinic acid and Salvia officinalis extracts on melanogenesis of B16F10 cells
Oliveira KB, et al.
Revista Brasileira de Farmacognosia, 23(2), 249-258 (2013)
Sunil Kalia et al.
Pigment cell & melanoma research, 31(1), 31-38 (2017-08-15)
Objective measurements of melanin can provide important information for differentiating melanoma from benign pigmented lesions and in assessing pigmentary diseases. Herein, we evaluate near-infrared (NIR) fluorescence as a possible tool to quantify melanin. Various concentrations of in vitro Sepia melanin
Detection of antibodies against Paracoccidioides brasiliensis melanin in in vitro and in vivo studies during infection.
Uran ME
Clinical and Vaccine Immunology : CVI, 18, 1680-1680 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service