M1180
Mineral oil
meets USP testing specifications
Synonym(s):
White mineral oil (petroleum)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
USP/NF
meets USP testing specifications
Quality Level
form
liquid
refractive index
n20/D 1.467 (lit.)
density
0.81-0.89 g/cm3 at 15 °C
application(s)
pharmaceutical (small molecule)
InChI key
AEOVEGJBKQQFOP-DDVLFWKVSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Observation of neutron emission during acoustic cavitation of deuterated titanium powder.: This study explores the novel use of mineral oil as a neutron moderator in experimental setups involving acoustic cavitation, highlighting its unique properties that aid in neutron capture and measurement in nuclear research (Fomitchev-Zamilov, 2024).
- Air trap and removal on a pressure-driven PDMS-based microfluidic device.: Demonstrating the utility of mineral oil in microfluidic applications, this research focuses on its role in managing air traps in lab-on-a-chip systems, crucial for biomedical and diagnostic device efficiency (Xu et al., 2024).
- Kinetic Assessment of Kraft and Thermally Upgraded Kraft Papers Aged in Five Alternative Dielectric Fluids.: Investigating the impact of mineral oil and alternative fluids on the aging behavior of kraft paper, this study contributes to better understanding of material durability in electrical insulation systems (Mendez et al., 2024).
Biochem/physiol Actions
Mineral oil functions by forming a depot at the injection site slowly releasing antigen.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 6(9), e24910-e24910 (2011-09-29)
Aptamers are short RNA or DNA oligonucleotides which can bind with different targets. Typically, they are selected from a large number of random DNA sequence libraries. The main strategy to obtain aptamers is systematic evolution of ligands by exponential enrichment
Clinical immunology (Orlando, Fla.), 127(1), 98-106 (2008-02-16)
The pathogenesis of T cell-mediated diseases like rheumatoid arthritis (RA) has typically been explained in the context of the Th1-Th2 paradigm: the initiation/propagation by pro-inflammatory cytokines, and downregulation by Th2 cytokines. However, in our study based on the adjuvant-induced arthritis
PloS one, 3(9), e3241-e3241 (2008-09-20)
There is increasing evidence that tissue-specific modifications of basic cellular functions play an important role in development and disease. To identify the functions of COPI coatomer-mediated membrane trafficking in Drosophila development, we were aiming to create loss-of-function mutations in the
PloS one, 6(10), e26443-e26443 (2011-10-27)
In nature the aerial trace of pheromone used by male moths to find a female appears as a train of discontinuous pulses separated by gaps among a complex odorant background constituted of plant volatiles. We investigated the effect of such
Nature methods, 6(2), 167-172 (2009-01-20)
Accurate chromosome segregation during meiosis depends on the assembly of a microtubule-based spindle of proper shape and size. Current models for spindle-size control focus on reaction diffusion-based chemical regulation and balance in activities of motor proteins. Although several molecular perturbations
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service