Skip to Content
Merck
All Photos(1)

Key Documents

L0750000

Loperamide hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H33ClN2O2 · HCl
CAS Number:
Molecular Weight:
513.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

loperamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4

InChI

1S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34H,17-22H2,1-2H3;1H

InChI key

PGYPOBZJRVSMDS-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Loperamide hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Natalja Genina et al.
International journal of pharmaceutics, 453(2), 488-497 (2013-06-19)
The primary goal of the current work was to study the applicability of precision inkjet printing in fabrication of personalized doses of active pharmaceutical ingredients (APIs). Loperamide hydrochloride (LOP) and caffeine (CAF) were used as model compounds. Different doses of
James Church et al.
Diseases of the colon and rectum, 56(1), 120-125 (2012-12-12)
Coating of the cecum with sticky bile causes a problem with inspection of the mucosa during colonoscopy. This study aimed to see whether 4 mg of loperamide taken after colonoscopy preparation would delay the passage of bile and improve the
Acute infectious diarrhea in immunocompetent adults.
Herbert L DuPont
The New England journal of medicine, 370(16), 1532-1540 (2014-04-18)
Linda Lefièvre et al.
The Biochemical journal, 448(2), 189-200 (2012-09-05)
Ca2+i signalling is pivotal to sperm function. Progesterone, the best-characterized agonist of human sperm Ca2+i signalling, stimulates a biphasic [Ca2+]i rise, comprising a transient and subsequent sustained phase. In accordance with recent reports that progesterone directly activates CatSper, the [Ca2+]i
E Placidi et al.
Alimentary pharmacology & therapeutics, 36(1), 64-73 (2012-05-16)
Loperamide (LOP) is an anti-diarrhoeal agent which is thought to act largely by slowing transit with an uncertain effect on the fluid content of the small and large bowel in humans. Adding simethicone (SIM) to LOP improves its efficacy, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service