Skip to Content
Merck
All Photos(2)

Key Documents

K1512

Sigma-Aldrich

Kaolin

meets USP testing specifications

Synonym(s):

China clay, Aluminum silicate hydroxide, Bolus, Hydrated aluminum silicate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
~Al2Si2O5(OH)4
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11111803
NACRES:
NA.25
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Quality Level

Agency

USP/NF
meets USP testing specifications

form

solid

application(s)

pharmaceutical (small molecule)

SMILES string

[Si](=O)(O[Si](=O)O[Al]=O)O[Al]=O.O.O

InChI

1S/2Al.O5Si2.2H2O.2O/c;;1-6(2)5-7(3)4;;;;/h;;;2*1H2;;/q2*+1;-2;;;;

InChI key

NLYAJNPCOHFWQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • A kaolin/calcium incorporated shape memory and antimicrobial chitosan-dextran based cryogel as an efficient haemostatic dressing for uncontrolled hemorrhagic wounds: Explores the use of kaolin in creating an innovative cryogel that offers improved haemostatic performance, crucial for managing severe bleeding in clinical settings (Andrabi and Kumar, 2023).

  • Properties of hydric and biodegradability of cassava starch-based bioplastics reinforced with thermally modified kaolin: Examines the enhancement of bioplastic properties through the incorporation of kaolin, aiming to improve water resistance and biodegradability, which is significant for environmental sustainability (Meite et al., 2021).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oded Pinkas et al.
Acta biomaterialia, 51, 125-137 (2017-01-23)
Bioadhesives are polymeric hydrogels that can adhere to a tissue after crosslinking and are an essential element in nearly all surgeries worldwide. Several bioadhesives are commercially available. However, none of them are ideal. The main limitation of current tissue adhesives
Kenji Inaba et al.
The journal of trauma and acute care surgery, 74(2), 538-545 (2013-01-29)
The purpose of this study was to evaluate the long-term efficacy and safety of kaolin- and chitosan-based hemostatic agents for hemorrhage control in a 14-day survival, damage-control swine model of Grade IV liver injury. A total of 48 anesthetized pigs
Maya Aleshnick et al.
The journal of trauma and acute care surgery, 76(1), 107-113 (2013-12-26)
Viscoelastic measurements are frequently being used in clinical and research settings for a rapid assessment of the hemostatic processes of blood clot formation and degradation. These measurements are being performed on either of two instruments (TEG and ROTEM) using their
Marta Markiewicz et al.
Chemosphere, 90(2), 706-712 (2012-10-20)
As the amount of industrial processes involving ionic liquids (ILs) increase the question of their environmental fate awaits an answer. Should ILs become a source of pollution they will primarily be found in soils and water. Interaction of imidazolium IL
Ahmad H Rajab Aljuboori et al.
Bioresource technology, 127, 489-493 (2012-11-20)
The production and characterization of a bioflocculant, IH-7, by Aspergillus flavus was investigated. About 0.4 g of purified bioflocculant with an average molecular weight of 2.574 × 10(4)Da could be obtained from 1L of fermentation medium. The bioflocculant mainly consisted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service