I0775020
Isosorbide 2-nitrate
European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
isosorbide
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
InChI
1S/C6H9NO6/c8-3-1-11-6-4(13-7(9)10)2-12-5(3)6/h3-6,8H,1-2H2/t3-,4+,5-,6-/m1/s1
InChI key
YWXYYJSYQOXTPL-JGWLITMVSA-N
Looking for similar products? Visit Product Comparison Guide
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Isosorbide 2-nitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Packaging
The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.
Other Notes
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pharmaceutical research, 10(1), 22-27 (1993-01-01)
The pharmacokinetics and urinary recoveries of four isomeric mononitrates, L-isoidide mononitrate (L-IIMN), isosorbide-2-mononitrate (IS-2-MN), isomannide mononitrate (IMMN), and isosorbide-5-mononitrate (IS-5-MN), were investigated at an intravenous dose of 2 mg/kg in rats. All four compounds exhibited monoexponential kinetics at this dose.
Acta medica Okayama, 44(1), 51-53 (1990-02-01)
The plasma level of isosorbide dinitrate intraperitoneally administered to rats stressed by foot-shock was almost the same as that in non-stressed control rats. However, levels of its metabolites, 5-isosorbide mononitrate and 2-isosorbide mononitrate, were lower in stressed rats than in
European journal of clinical pharmacology, 38(2), 145-147 (1990-01-01)
The concentrations of isosorbide dinitrate (ISDN), isosorbide-5-mononitrate (IS-5-MN) and isosorbide-2-mononitrate (IS-2-MN) were determined in plasma (PL), saphenous vein wall (SV) and pectoral muscle (PM) from 8 patients undergoing coronary bypass surgery. The patients were pretreated for 2 days with ISDN
Masui. The Japanese journal of anesthesiology, 42(2), 225-232 (1993-02-01)
Nineteen patients with ischemic heart disease were studied to determine plasma levels of isosorbide dinitrate (ISDN) and its metabolites, isosorbide-2-mononitrate (2-ISMN) and isosorbide-5-mononitrate (5-ISMN) for 6 hrs during intravenous administration of ISDN, using gas chromatography. Differences in plasma levels of
Drug metabolism and disposition: the biological fate of chemicals, 18(4), 429-434 (1990-07-01)
A pharmacokinetic model is proposed to describe the plasma levels of isosorbidedinitrate (ISDN) and its two pharmacologically active metabolites, isosorbide-2-mononitrate (IS-2MN) and isosorbide-5-mononitrate (IS-5MN), following the oral administration of several 20-mg sustained release formulations of ISDN. Absorption of ISDN from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service