Skip to Content
Merck
All Photos(1)

Key Documents

I0060000

Ifosfamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ifex, N,3-Bis(2-chloroethyl)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine-2-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O2P
CAS Number:
Molecular Weight:
261.09
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

ifosfamide

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

ClCCNP1(=O)OCCCN1CCCl

InChI

1S/C7H15Cl2N2O2P/c8-2-4-10-14(12)11(6-3-9)5-1-7-13-14/h1-7H2,(H,10,12)

InChI key

HOMGKSMUEGBAAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ifosfamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Ifosfamide is a nitrogen mustard compound that is a structural isomer of cyclophosphamide. Ifosfamide is a prodrug that must be transformed by cytochrome P450 to the biologically active component. It is used as an antineoplastic agent in cancer chemotherapy, but ifosfamide is more likely to cause renal toxicity than cyclophosphamide.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metin Tascilar et al.
The oncologist, 12(11), 1351-1360 (2007-12-07)
The treatment outcome of patients with locally advanced and metastatic soft tissue sarcomas is poor. Doxorubicin is regarded as standard treatment, but its use is featured by the occurrence of cardiotoxicity. This hinders the administration of this drug at high
T Ajithkumar et al.
Clinical oncology (Royal College of Radiologists (Great Britain)), 19(2), 108-114 (2007-03-16)
Encephalopathy is a potentially fatal toxicity of ifosfamide. Clinical manifestations of encephalopathy range from fatigue and confusion to coma and death. Early identification of this toxicity and prompt cessation of ifosfamide are the essential elements in the management of ifosfamide
Jean-Yves Blay et al.
European journal of cancer (Oxford, England : 1990), 50(6), 1137-1147 (2014-02-12)
This randomised phase III trial evaluated first-line trabectedin versus doxorubicin-based chemotherapy (DXCT) in patients with advanced/metastatic translocation-related sarcomas (TRS). Patients were randomly assigned (1:1) to receive trabectedin 1.5mg/m2 24-h intravenous (i.v.) infusion every 3 weeks (q3wk) (Arm A), or doxorubicin
Renée L Mulder et al.
The Cochrane database of systematic reviews, (2)(2), CD006300-CD006300 (2010-02-19)
Alkylating agents, such as cyclophosphamide and ifosfamide, play a major role in the improved survival of children and young adults with bone and soft tissue sarcoma. However, there is still controversy as to their comparative anti-tumour efficacy and possible adverse
Yesne Alici-Evcimen et al.
Psycho-oncology, 16(10), 956-960 (2007-02-06)
Ifosfamide is a chemotherapy agent commonly used in the treatment of several solid tumors and hematologic malignancies. Common ifosfamide toxicities are bone marrow suppression, alopecia, nausea and vomiting. Less common, but striking, are the rare but dramatic neuropsychiatric toxicities with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service