Skip to Content
Merck
All Photos(1)

Key Documents

H0800000

Homatropine hydrobromide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

DL-Homatropine hydrobromide, DL-endo-α-Hydroxybenzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester hydrobromide, Tropine mandelate hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO3 · HBr
CAS Number:
Molecular Weight:
356.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

homatropine

manufacturer/tradename

EDQM

mp

214-217 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Br.CN1C2CCC1CC(C2)OC(=O)C(O)c3ccccc3

InChI

1S/C16H21NO3.BrH/c1-17-12-7-8-13(17)10-14(9-12)20-16(19)15(18)11-5-3-2-4-6-11;/h2-6,12-15,18H,7-10H2,1H3;1H

InChI key

DWSGTFTVBLXELC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Homatropine hydrobromide is an anticholinergic agent and is commonly used as a mydriatic and cycloplegic drug in ophthalmology.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amol Tamhane et al.
Ophthalmology, 112(11), 1963-1969 (2005-10-04)
To evaluate the role of amniotic membrane transplantation (AMT) in acute ocular burns. Prospective, randomized, controlled clinical trial. Patients with grade II to IV ocular burns within 3 weeks of injury were recruited. Thirty-seven patients, 7 of whom had bilateral
Elizabeth Hanno et al.
Optometry (St. Louis, Mo.), 80(2), 70-75 (2009-02-04)
Bilateral, recurrent, or chronic anterior uveitis requires a diagnostic evaluation to rule out any systemic cause. An understanding of the possible etiologies and their diagnostic criteria is needed to manage these patients. Treating any systemic cause can decrease the recurrent
Homatropine and psychosis in Weill-Marchesani syndrome.
Anjaiah Madugula et al.
Journal of the American Academy of Child and Adolescent Psychiatry, 45(7), 769-770 (2006-07-13)
Brian M Thome et al.
Journal of chromatography. A, 1138(1-2), 291-300 (2006-11-14)
Over the past decade the moving bed process has become a commonly used tool for the continuous separation of chiral compounds, and its recent application to electrophoretic separations allows the technique to be used as a model system for moving
Topical chloramphenicol prophylaxis is of dubious benefit in corneal abrasion.
Joseph Ting
Emergency medicine Australasia : EMA, 23(2), 231-231 (2011-04-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service