Skip to Content
Merck
All Photos(5)

Documents

F8761

Sigma-Aldrich

Fast Garnet GBC sulfate salt

Powder

Synonym(s):

2-Methyl-4-([2-methylphenyl]azo)benzenediazonium salt, 4-Amino-2′,3-dimethylazobenzene diazotated, Azoic Diazo No. 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O4S
CAS Number:
Molecular Weight:
334.35
Colour Index Number:
37210
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Fast Garnet GBC sulfate salt, diazonium dye

Quality Level

form

powder

composition

Dye Content Calculated, ≥90%

color

orange to very dark orange, and Orange-Brown to Very Dark Orange-Brown and Red to Very Dark

solubility

H2O: 5 mg/mL, clear

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

OS([O-])(=O)=O.Cc1ccccc1N=Nc2ccc([N+]#N)c(C)c2

InChI

1S/C14H13N4.H2O4S/c1-10-5-3-4-6-14(10)18-17-12-7-8-13(16-15)11(2)9-12;1-5(2,3)4/h3-9H,1-2H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

PPTXFSQSISUDAU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Fast Garnet GBC is a diazonium dye and is mainly required in azo-dye methods. In alkaline phosphatase staining, it reacts with α-naphthol to give a red insoluble azo dye at the site of enzyme activity.

Application

Fast Garnet GBC sulfate salt has been used for the staining of aspartate aminotransferase activity. It has been used for measuring of the non-specific esterase activity.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Incipient speciation across a depth gradient in a scleractinian coral?
Carlon DB and Budd AF
Evolution, 56, 2227-2227 (2002)
Evidence of multiple insecticide resistance mechanisms in Anopheles gambiae populations in Bangui, Central African Republic.
Ole Sangba ML et al.
Parasites & vectors, 10, 23-23 (2017)
Bancroft JD
Theory and Practice of Histological Techniques (2008)
Olga Liliana Anguiano et al.
Aquatic toxicology (Amsterdam, Netherlands), 188, 72-79 (2017-05-02)
During the last years, a carbaryl insecticide was extensively applied in the valley of Río Negro and Neuquén, North Patagonia Argentina, to manage codling moths (Cydia pomonella), the main pest of pear and apple trees. In this study carbaryl susceptibility
Steven J Brooks et al.
Aquatic toxicology (Amsterdam, Netherlands), 160, 1-12 (2015-01-09)
Mytilus species are important organisms in marine systems being highly abundant and widely distributed along the coast of Europe and worldwide. They are typically used in biological effects studies and have a suite of biological effects endpoints that are frequently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service