Skip to Content
Merck
All Photos(1)

Key Documents

E1900000

Ethinylestradiol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17α-Ethynylestradiol, 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Beilstein:
2419975
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

ethinyl estradiol

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

182-183 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@@]3(O)C#C)[C@]1([H])CCc4cc(O)ccc24

InChI

1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1

InChI key

BFPYWIDHMRZLRN-SLHNCBLASA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethinylestradiol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Han et al.
Water research, 47(7), 2273-2284 (2013-03-08)
This study demonstrates that ethinylestradiol (EE2), a priority estrogenic contaminant in water, can be rapidly and selectively removed from aqueous solutions using industrial-grade polyamide 612 (PA612) particles as adsorbents. Isothermal studies showed that nonporous low surface area (20 m(2) g(-1)) PA612 particles
Konrad F Koehler et al.
Endocrine reviews, 26(3), 465-478 (2005-04-29)
We have known for many years that estrogen is more than the female hormone. It is essential in the male gonads, and in both sexes, estrogen has functions in the skeleton and central nervous system, on behavior, and in the
C Piérard-Franchimont et al.
Maturitas, 22(2), 151-154 (1995-09-01)
The effects of menopause and hormone replacement therapy (HRT) on the epidermis are largely unknown. The aim of this study was to model and measure the influence of estrogen-transdermal HRT on subtle physiological changes taking place in the epidermis during
Bin Huang et al.
Environment international, 59, 262-273 (2013-07-16)
The occurrence, removal and bioaccumulation of steroid estrogens such as the natural estrone (E1), 17β-estradiol (E2) and estriol (E3), as well as the synthetic 17α-ethynylestradiol (EE2) were investigated in Dianchi Lake catchment, China. The results show that traditional secondary treatment
Jürgen Dinger et al.
Obstetrics and gynecology, 122(4), 800-808 (2013-10-03)
To compare the risks of short-term and long-term use of an etonogestrel-containing and ethinylestradiol-containing vaginal ring and combined oral contraceptive pills (OCPs) in a routine clinical study population. This was a prospective, controlled, noninterventional cohort study performed in the United

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service