Skip to Content
Merck
All Photos(1)

Key Documents

E1500000

Erythromycin ethyl succinate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H75NO16
CAS Number:
Molecular Weight:
862.05
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

erythromycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(=O)CCC(=O)OC1C(CC(C)OC1OC2C(C)C(OC3CC(C)(OC)C(O)C(C)O3)C(C)C(=O)OC(CC)C(C)(O)C(O)C(C)C(=O)C(C)CC2(C)O)N(C)C

InChI

1S/C43H75NO16/c1-15-29-43(11,52)36(48)24(5)33(47)22(3)20-41(9,51)38(25(6)34(26(7)39(50)57-29)59-32-21-42(10,53-14)37(49)27(8)56-32)60-40-35(28(44(12)13)19-23(4)55-40)58-31(46)18-17-30(45)54-16-2/h22-29,32,34-38,40,48-49,51-52H,15-21H2,1-14H3

InChI key

NSYZCCDSJNWWJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Erythromycin ethyl succinate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Brook et al.
Clinical therapeutics, 23(11), 1889-1900 (2002-01-05)
Cefprozil and erythromycin are acceptable alternatives to penicillin in the treatment of pharyngitis/tonsillitis due to group A beta-hemolytic streptococcus (GABHS). The purpose of this trial was to determine the relative efficacy and tolerability of cefprozil and erythromycin in the treatment
Yi Gu et al.
Journal of pharmaceutical and biomedical analysis, 40(3), 737-743 (2005-12-27)
A sensitive and selective liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS) method was developed for simultaneous identification and quantification of erythromycin ethylsuccinate and erythromycin in human plasma, which can be well applied to clinical study. The method was based on liquid-liquid
J J Stewart et al.
Journal of clinical pharmacology, 40(4), 347-353 (2000-04-13)
Motion sickness and the antimotion sickness drugs scopolamine (SCP) and promethazine (PMZ) inhibit gastric emptying (GE). This study was conducted to determine if erythromycin would exert its well-known prokinetic effects in normal and motion-sick subjects given antimotion sickness drugs. Fifteen
Y A Cho et al.
Die Pharmazie, 67(2), 124-130 (2012-04-20)
The effects of curcumin, a natural anti-cancer compound, on the bioavailability and pharmacokinetics of tamoxifen and its metabolite, 4-hydroxytamoxifen, were investigated in rats. Tamoxifen and curcumin interact with cytochrom P450 (CYP) enzymes and P-glycoprotein, and the increase in the use
Yan-Chun Feng et al.
Journal of pharmaceutical and biomedical analysis, 41(2), 373-384 (2006-01-13)
Universal quantitative models using NIR reflectance spectroscopy were developed for the analysis of API contents (active pharmaceutical ingredient) in roxithromycin and erythromycin ethylsuccinate tablets from different manufacturers in China. The two quantitative models were built from 78 batches of roxithromycin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service