Skip to Content
Merck
All Photos(1)

Key Documents

D9516

Supelco

Diaveridine

analytical standard, ≥99.0% (TLC)

Synonym(s):

5-(3,4-Dimethoxybenzyl)-2,4-pyrimidinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N4O2
CAS Number:
Molecular Weight:
260.29
Beilstein:
258464
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (TLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(Cc2cnc(N)nc2N)cc1OC

InChI

1S/C13H16N4O2/c1-18-10-4-3-8(6-11(10)19-2)5-9-7-16-13(15)17-12(9)14/h3-4,6-7H,5H2,1-2H3,(H4,14,15,16,17)

InChI key

LDBTVAXGKYIFHO-UHFFFAOYSA-N

Gene Information

rat ... Dhfr(24312)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Residue Reviews: Residues of Pesticides and Other Contaminants in the Total Environment (2012)
H Yoshimura
Mutation research, 261(2), 149-152 (1991-10-01)
The coccidiostat diaveridine was tested for mutagenicity in the Salmonella/microsome assay with tester strains TA100 and TA98. This compound was not mutagenic in either tester strain in the presence and absence of rat S9 mix, but was found to be
P Tömpe et al.
Acta pharmaceutica Hungarica, 60(1), 23-29 (1990-01-01)
Differential pulse polarographic method was applied for determination of diaveridine in plasma, whole blood and isolated organs (liver, spleen, kidney, thymus). Deproteinization of homogenized blood and organs had been carried out on Extrelut column and then concentration of diaveridine was
A Tanabe et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 86(6), 411-415 (1985-12-01)
The preventive effect of sulfaquinoxaline and diaveridine in combination against leucocytozoon infection in White Leghorn female chickens was tested in a field trial. The experiment was initiated when the animals were 120 days old, and it lasted for 10 weeks.
Determination of residual diaveridine and sulfaquinoxaline in Hen's egg, chicken plasma and tissues by high-performance liquid chromatography.
T Sakano et al.
Chemical & pharmaceutical bulletin, 29(8), 2290-2295 (1981-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service