Skip to Content
Merck
All Photos(1)

Documents

D7282

Supelco

Desmethyldiazepam

analytical standard

Synonym(s):

Nordiazepam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClN2O
CAS Number:
Molecular Weight:
270.71
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to light tan

solubility

DMSO: >10 mg/mL

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

room temp

SMILES string

Clc1ccc2NC(=O)CN=C(c3ccccc3)c2c1

InChI

1S/C15H11ClN2O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9H2,(H,18,19)

InChI key

AKPLHCDWDRPJGD-UHFFFAOYSA-N

Gene Information

rat ... Tspo(24230)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Primary metabolite of diazepam; ligand for the GABAA receptor benzodiazepine modulatory site.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pi Cheng et al.
Bioorganic & medicinal chemistry letters, 18(13), 3787-3789 (2008-06-06)
A series of 4-aryl-6-chloro-quinolin-2-ones and 5-aryl-7-chloro-1,4-benzodiazepine were synthesized and assayed for their in vitro anti-hepatitis B virus activities and cytotoxicities for the first time. Some of the tested compounds were active against HBsAg and HBeAg secretion in Hep G2.2.15 cells.
Joris C Verster et al.
Sleep medicine reviews, 17(2), 153-159 (2012-08-14)
The use of benzodiazepine receptor agonists can significantly impair driving performance. The aim of this review was to determine if there is a relation between blood concentrations of these drugs and the degree of driving impairment. A literature search was
Laura Mercolini et al.
Talanta, 80(1), 279-285 (2009-09-29)
Diazepam is frequently used as an adjuvant during antidepressant therapy. Recently, some studies have suggested that the treatment with benzodiazepines could have different efficacy in depressed patients as opposed to non-depressed ones. To clarify the matter, a study is currently
Michael J Lamson et al.
Clinical drug investigation, 31(8), 585-597 (2011-07-05)
Acute repetitive seizures (ARS) are a debilitating part of episodic seizure activity that can sometimes progress to status epilepticus. Currently approved treatment that can be administered by non-medical personnel to patients with ARS is a diazepam rectal gel. While effective
Ali Acikgöz et al.
Toxicology and applied pharmacology, 234(2), 179-191 (2008-11-06)
Drug biotransformation is one of the most important parameters of preclinical screening tests for the registration of new drug candidates. Conventional existing tests rely on nonhuman models which deliver an incomplete metabolic profile of drugs due to the lack of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service