Skip to Content
Merck
All Photos(1)

Key Documents

D1039990

Diclofenac potassium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Diclofenac potassium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2KNO2
CAS Number:
Molecular Weight:
334.24
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

diclofenac

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[K+].Clc1c(c(ccc1)Cl)Nc2c(cccc2)CC(=O)[O-]

InChI

1S/C14H11Cl2NO2.K/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1

InChI key

KXZOIWWTXOCYKR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diclofenac potassium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

W McNeely et al.
Drugs, 57(6), 991-1003 (1999-07-10)
The NSAID diclofenac is a potent inhibitor of prostaglandin synthesis and an established antipyretic and analgesic agent. Diclofenac-potassium was developed as an immediate-release tablet with the aim of providing rapid onset of action after oral administration. This formulation has been
Nicholas Moore
Clinical drug investigation, 27(3), 163-195 (2007-02-20)
Non-prescription (over-the-counter [OTC]) analgesics are used for the short-term treatment of acute painful conditions of mild to moderate intensity in everyday life. Well documented safety and efficacy, a rapid onset of action and a flexible daily dosing regimen are essential
Seham Mustafa et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(5), 801-806 (2013-03-27)
Renal function and disease are commonly evaluated by radionuclide studies. The choice of radiopharmaceutical agent for various studies is crucial for proper interpretation. (99m)Tc-mercaptoacetyltriglycine ((99m)Tc-MAG3) is excreted almost exclusively by the renal tubules, whereas (99m)Tc-diethylenetriamine pentaacetic acid ((99m)Tc-DTPA) is predominantly
Rachel P L van Swelm et al.
Toxicology and applied pharmacology, 269(2), 141-149 (2013-03-20)
Diclofenac (DF) is a widely used non-steroidal anti-inflammatory drug for the treatment of rheumatic disorders, but is often associated with liver injury. We applied urinary proteomic profiling using MALDI-TOF MS to identify biomarkers for DF-induced hepatotoxicity in mice. Female CH3/HeOUJIco
Stanislava Stepanova et al.
Environmental toxicology and pharmacology, 35(3), 454-460 (2013-03-19)
Diclofenac residues have been found in surface water, and thus could present a potential risk to aquatic species. The aim of this study was to assess the impact of diclofenac on the mortality, growth, and development of fish, as well

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service