Skip to Content
Merck
All Photos(1)

Key Documents

CRMUDEGO

Density Standard

ASTM D 4052; IP 365 / ISO 12185 (0.8355g/mL, 15 °C)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

according to ASTM® D 4052

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM) certified to ISO 17034 and ISO/IEC 17025 under UKAS accreditation. The certified value of the CRM is determined with the use of a digital density meter.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Density Standards, ASTM D 4052 can be found here.

Application

Density measurement ASTM standard for routine verification of density meters.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service