Skip to Content
Merck
All Photos(5)

Key Documents

C8532

Sigma-Aldrich

Sodium citrate tribasic dihydrate

for molecular biology, ≥99%

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

Molecular Biology
for molecular biology

Quality Level

Assay

≥99%

form

powder

pH

7.5-9 (25 °C, 29.4 g/L)

mp

>300 °C (lit.)

solubility

water: soluble 100 mg/mL

foreign activity

DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
Protease, none detected
RNAse, none detected

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Sodium citrate tribasic dihydrate is a stabilizer that imparts colloidal stability to gold nanoparticles (AuNPs).

Application

Sodium citrate tribasic dihydrate has been used:
  • to perform antibody retrieval for the immunohistochemical analysis of ovine ovarian sections
  • as a component of induction medium to culture Agrobacterium strain C58C1
  • to isolate human neutrophils from whole blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Lalonde et al.
Genome biology, 20(1), 133-133 (2019-07-10)
Genome-wide association studies (GWAS) have identified hundreds of loci associated with coronary artery disease (CAD) and blood pressure (BP) or hypertension. Many of these loci are not linked to traditional risk factors, nor do they include obvious candidate genes, complicating
Enrique Gómez et al.
Theriogenology, 146, 39-47 (2020-02-10)
Direct transfer (DT) of cryopreserved embryos to recipients facilitates on-farm application. We analyzed a new freezing/thawing (F/T) procedure for in vitro produced (IVP) embryos, integrating: 1) an ethylene-glycol based system; 2) a culture step without protein; and 3) a synthetic protein
Fengyu Zhang et al.
BMC cancer, 20(1), 673-673 (2020-07-20)
Glioblastoma is devastating cancer with a high frequency of occurrence and poor survival rate and it is urgent to discover novel glioblastoma-specific antigens for the therapy. Cancer-germline genes are known to be related to the formation and progression of several
Kwi Hye Koh et al.
The Journal of clinical investigation, 129(4), 1713-1726 (2019-02-13)
Soluble urokinase receptor (suPAR) is a circulatory molecule that activates αvβ3 integrin on podocytes, causes foot process effacement, and contributes to proteinuric kidney disease. While active integrin can be targeted by antibodies and small molecules, endogenous inhibitors haven't been discovered
Mike R Wilson et al.
Cell reports, 33(6), 108366-108366 (2020-11-12)
Endometriosis affects 1 in 10 women and is characterized by the presence of abnormal endometrium at ectopic sites. ARID1A mutations are observed in deeply invasive forms of the disease, often correlating with malignancy. To identify epigenetic dependencies driving invasion, we

Protocols

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service