Skip to Content
Merck
All Photos(1)

Documents

C2400000

Clonidine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(2,6-Dichloroanilino)-2-imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9Cl2N3 · HCl
CAS Number:
Molecular Weight:
266.55
Beilstein:
4163525
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clonidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Clc1cccc(Cl)c1NC2=NCCN2

InChI

1S/C9H9Cl2N3.ClH/c10-6-2-1-3-7(11)8(6)14-9-12-4-5-13-9;/h1-3H,4-5H2,(H2,12,13,14);1H

InChI key

ZNIFSRGNXRYGHF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clonidine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula E Park et al.
Addiction biology, 20(2), 275-284 (2013-12-18)
Opioids represent effective drugs for the relief of pain, yet chronic opioid use often leads to a state of increased sensitivity to pain that is exacerbated during withdrawal. A sensitization of pain-related negative affect has been hypothesized to closely interact
Kalpashri Kesavan et al.
Respiratory physiology & neurobiology, 201, 93-100 (2014-07-11)
Sedative-analgesics are often given to newborn infants and are known to affect many components of the autonomic nervous system. While morphine is most frequently used, α-2 adrenergic receptor agonists are being increasingly used in this population. Alpha-2 adrenergic receptors agonists
Craige C Wrenn et al.
Neuroscience letters, 585, 109-113 (2014-11-30)
Fragile X syndrome (FXS), a disorder caused by a mutation in the FMR1 gene, is often associated with Attention Deficit Hyperactivity Disorder (ADHD). Common treatments for the hyperactivity often seen in ADHD involve the use of stimulants and α2-adrenergic agonists.
Yanyan Qiu et al.
Scientific reports, 4, 6605-6605 (2014-10-14)
This study examined whether a novel imidazoline I₂ receptor ligand CR4056 could serve as a discriminative stimulus and whether it shares similar discriminative stimulus effects with other reported I₂ receptor ligands. Eight male Sprague-Dawley rats were trained to discriminate 10.0 mg/kg
Jonathan W Pinkston et al.
Behavioural pharmacology, 25(7), 629-636 (2014-08-15)
Under many circumstances, reinforcer magnitude appears to modulate the rate-dependent effects of drugs such that when schedules arrange for relatively larger reinforcer magnitudes rate dependency is attenuated compared with behavior maintained by smaller magnitudes. The current literature on resistance to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service