Skip to Content
Merck
All Photos(1)

Key Documents

C1795

Sigma-Aldrich

Canada balsam

Viscous Liquid

Synonym(s):

Balsam Canada

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Canada balsam, Mounting medium for microscopy

Quality Level

form

viscous liquid

technique(s)

microbe id | staining: suitable

color

yellow to very dark yellow

refractive index

n20/D 1.522 (lit.)

density

0.99 g/mL

application(s)

hematology
histology

storage temp.

room temp

SMILES string

CC=CC=C(C=C)C(CN1CCC(NC(C1=O)CCCN=C(N)N)C(CC2=CC3=CC=CC=C3C=C2)NC(=O)C)C(=CC=C)C=CC

InChI

InChI=1S/C39H52N6O2/c1-6-10-16-30(9-4)34(32(14-7-2)15-8-3)27-45-24-22-35(44-36(38(45)47)19-13-23-42-39(40)41)37(43-28(5)46)26-29-20-21-31-17-11-12-18-33(31)25-29/h6-12,14-18,20-21,25,34-37,44H,2,4,13,19,22-24,26-27H2,1,3,5H3,(H,43,46)(H4,40,41,42)

InChI key

IIUZTTUQXFCORF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Canada Balsam is a natural mounting medium obtained from balsam fir trees. It is produced from the resin of the balsam fir tree and its use can be combined with xylene-containing specimens. The optical properties of Canada Balsam are nearly identical to those of glass. Specimens must be dehydrated before using this medium. Permanent slides mounted with Canada Balsam have been stored for over a century.

Application

Canada balsam is used as a stable apolar hydrophobic mounting medium for light microscopy slide preparation, especially when long-term storage is desired. It is mostly applicable for cellular staining.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chick A
Insect Microscopy (2016)
Natalija Filipovic et al.
Histochemistry and cell biology, 147(4), 481-495 (2016-10-04)
Differentiation of human podocytes starts with mesenchymal-to-epithelial transition (MET) of the metanephric mesenchyme into the S-shaped nephrons. During further development, differentiating podocytes regain mesenchyme-like cell characteristics by epithelial-to-mesenchymal transition (EMT), leading to formation of the terminally differentiated, non-dividing cell. Both
Identification of carotid 'vulnerable plaque' by contrast-enhanced ultrasonography: correlation with plaque histology, symptoms and cerebral computed tomography.
Faggioli GL, et al.
European Journal of Vascular and Endovascular Surgery, 41, 238-238 (2011)
Activation of caspases and p53 by bovine herpesvirus 1 infection results in programmed cell death and efficient virus release.
Devireddy LR and Jones CJ
Journal of Virology, 73, 3778-3778 (1999)
Vlatka Martinovic et al.
Anatomical record (Hoboken, N.J. : 2007), 300(7), 1315-1326 (2016-12-17)
Expressions of cytokeratin 8 (CK8), vimentin, nestin, and alpha-smooth-muscle-actin (alpha-SMA) were analyzed in the developing gonads of 12, 5-9 week old (W) human conceptuses by immunohistochemistry and immunofluorescence. During the investigated period, the number of CK8 positive cells increased from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service